Orders LinkContactUsLinkComponent
Metabolomics column kits
Metabolomics column kits
Supplier:   MICROSOLV
Metabolomics column kits
Supplier:  MICROSOLV
Certificates
Metabolomics column kits
Metabolomics column kits
6 Options Available Below
Product Details & Documents
Each kit includes one Cogent Bidentate C18™ column and one Cogent Diamond Hydride™ column.
Product Family Options
Product Information
  • Column Format
    Phase
    Pore size
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • Analytical
    Diamond hydride and Bidentate C18
    100 Å
    4 µm
    2,10 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 1 490,00 €
    1 490,00 €
    1 * 2 ST
       1 490,00 €
  • Analytical
    Diamond hydride and Bidentate C18
    100 Å
    4 µm
    4,60 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 1 770,00 €
    1 770,00 €
    1 * 2 ST
       1 770,00 €
  • Capillary
    Diamond hydride and Bidentate C18
    100 Å
    4 µm
    4,60 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 3 170,00 €
    3 170,00 €
    1 * 2 ST
       3 170,00 €
  • Analytical
    Diamond hydride and Bidentate C18
    100 Å
    4 µm
    2,10 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 1 760,00 €
    1 760,00 €
    1 * 2 ST
       1 760,00 €
  • Analytical
    Diamond hydride and Bidentate C18
    100 Å
    4 µm
    4,60 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 1 990,00 €
    1 990,00 €
    1 * 2 ST
       1 990,00 €
  • Analytical
    Diamond hydride and Bidentate C18
    100 Å
    4 µm
    2,10 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 1 980,00 €
    1 980,00 €
    1 * 2 ST
       1 980,00 €