

Formaldehyde 4%, pink, Q Path®, buffered
Supplier: VWR Collection
Certificates
About this item
CAS Number:50-00-0
:
Formula:CH₂O
MDL Number:MFCD00003274
:30.03 g/mol
Density:1.013 g/cm³ (20 °C)
EINECS:200-001-8
:GHS08
:GHS07
Storage Temperature:Ambient
:1
Specification Test Results
- Appearance:Pink liquid
- Test formaldehyde:Passes test
- Density (20/4):1,012 - 1,014
- pH (20°C):6,8 - 7,2
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRSFOR0024A910014 spec_VWRSFOR0024AB10141 spec_VWRSFOR0064A060014
Product specification
Frequently Bought Together