Orders LinkContactUsLinkComponent
96-Well plate sample control system, U-2D™
96-Well plate sample control system, U-2D™
Supplier:   MICROSOLV
96-Well plate sample control system, U-2D™
Supplier:  MICROSOLV
Certificates
96-Well plate sample control system, U-2D™
96-Well plate sample control system, U-2D™
31 Options Available Below
Product Details & Documents
The U-2D™ universal two dimensional 96-well plate system offer users unprecedented sample management control compared to current glass lined or plastic well plates for sample preparation or chromatography.
Easily push up the insert from the bottom and remove it from the rack. These inserts can then be placed into any MicroSolv 9 mm screw top autosampler vial for safe storage, transport or for a second chromatography dimension, or to another instrument platform.
Ordering information: Base and sealing mats sold separately.
Product Family Options
Inserts
Product Information
  • Availability
    Price
  • MICS95296-05ND
    Inserts, clear RSA-pro glass, 350 µl, for use with U-2D™ rack
    Inserts, clear RSA-pro glass, 350 µl, for use with U-2D™ rack
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 194,00 €
    194,00 €
    1 * 100 ST
       194,00 €
  • MICS95296-05N
    Inserts, clear borocilicate glass, 350 µl, for use with U-2D™ rack
    Inserts, clear borocilicate glass, 350 µl, for use with U-2D™ rack
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 125,00 €
    125,00 €
    1 * 100 ST
       125,00 €
  • MICS95296-I-1ML
    Inserts, clear borocilicate glass, 1 ml, for use with U-2D™ rack
    Inserts, clear borocilicate glass, 1 ml, for use with U-2D™ rack
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 136,00 €
    136,00 €
    1 * 100 ST
       136,00 €
  • MICS95296-I-1MLMR
    Inserts, clear RSA-pro glass, 1 ml, for use with U-2D™ rack
    Inserts, clear RSA-pro glass, 1 ml, for use with U-2D™ rack
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 207,00 €
    207,00 €
    1 * 100 ST
       207,00 €
  • MICS95296-I-500
    Inserts, clear borocilicate glass, 500 µl, for use with U-2D™ rack
    Inserts, clear borocilicate glass, 500 µl, for use with U-2D™ rack
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 114,00 €
    114,00 €
    1 * 100 ST
       114,00 €
  • MICS95296-I-500MR
    Inserts, clear RSA-pro glass, 500 µl, for use with U-2D™ rack
    Inserts, clear RSA-pro glass, 500 µl, for use with U-2D™ rack
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 194,00 €
    194,00 €
    1 * 100 ST
       194,00 €
Easy purchase pack U-2D™ plates
Product Information
  • Availability
    Price
  • MICS95296-1ML-1P
    U-2D™ rack with 96 well plate, 1 ml preloaded inserts, base and plug-style non-slit silicone sealing mat
    U-2D™ rack with 96 well plate, 1 ml preloaded inserts, base and plug-style non-slit silicone sealing mat
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 134,00 €
    134,00 €
    1 * 1 ST
       134,00 €
  • MICS95296-1ML-1R
    U-2D™ rack with 96 well plate, 1 ml preloaded inserts, base and plug-style non-slit silicone sealing mat
    U-2D™ rack with 96 well plate, 1 ml preloaded inserts, base and plug-style non-slit silicone sealing mat
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 152,00 €
    152,00 €
    1 * 1 ST
       152,00 €
  • MICS95296-1ML-3P
    U-2D™ rack with 96 well plate, 1 ml preloaded inserts, base and plug-style pre-slit silicone sealing mat
    U-2D™ rack with 96 well plate, 1 ml preloaded inserts, base and plug-style pre-slit silicone sealing mat
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 137,00 €
    137,00 €
    1 * 1 ST
       137,00 €
  • MICS95296-1ML-3R
    U-2D™ rack with 96 well plate, 1 ml preloaded inserts, base and chemical resistant pre-slit mat
    U-2D™ rack with 96 well plate, 1 ml preloaded inserts, base and chemical resistant pre-slit mat
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 156,00 €
    156,00 €
    1 * 1 ST
       156,00 €
  • MICS95296-1ML-PK
    U-2D™ rack with 96 well plate, 350 µl inserts preloaded
    U-2D™ rack with 96 well plate, 350 µl inserts preloaded
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 760,00 €
    760,00 €
    1 * 6 ST
       760,00 €
  • MICS95296-350-1DP
    U-2D™ rack with 96 well plate, 350 µl preloaded inserts, base and plug-style non-slit silicone sealing mat
    U-2D™ rack with 96 well plate, 350 µl preloaded inserts, base and plug-style non-slit silicone sealing mat
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 149,00 €
    149,00 €
    1 * 1 ST
       149,00 €
  • MICS95296-350-1DR
    U-2D™ rack with 96 well plate, 350 µl preloaded inserts, base and plug-style non-slit silicone sealing mat
    U-2D™ rack with 96 well plate, 350 µl preloaded inserts, base and plug-style non-slit silicone sealing mat
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 177,00 €
    177,00 €
    1 * 1 ST
       177,00 €
  • MICS95296-350-1P
    U-2D™ rack with 96 well plate, 350 µl preloaded inserts, base and plug-style non-slit silicone sealing mat
    U-2D™ rack with 96 well plate, 350 µl preloaded inserts, base and plug-style non-slit silicone sealing mat
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 134,00 €
    134,00 €
    1 * 1 ST
       134,00 €
  • MICS95296-350-1R
    U-2D™ rack with 96 well plate, 350 µl preloaded inserts, base and chemical resistant non-slit mat
    U-2D™ rack with 96 well plate, 350 µl preloaded inserts, base and chemical resistant non-slit mat
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 128,00 €
    128,00 €
    1 * 1 ST
       128,00 €
  • MICS95296-350-3DP
    U-2D™ rack with 96 well plate, 350 µl preloaded inserts, base and plug-style non-slit silicone sealing mat
    U-2D™ rack with 96 well plate, 350 µl preloaded inserts, base and plug-style non-slit silicone sealing mat
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 155,00 €
    155,00 €
    1 * 1 ST
       155,00 €
  • MICS95296-350-3DR
    U-2D™ rack with 96 well plate, 350 µl preloaded inserts, base and plug-style non-slit silicone sealing mat
    U-2D™ rack with 96 well plate, 350 µl preloaded inserts, base and plug-style non-slit silicone sealing mat
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 181,00 €
    181,00 €
    1 * 1 ST
       181,00 €
  • MICS95296-350-3P
    U-2D™ rack with 96 well plate, 350 µl preloaded inserts, base and plug-style non-slit silicone sealing mat
    U-2D™ rack with 96 well plate, 350 µl preloaded inserts, base and plug-style non-slit silicone sealing mat
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 138,00 €
    138,00 €
    1 * 1 ST
       138,00 €
  • MICS95296-350-3R
    U-2D™ rack with 96 well plate, 350 µl preloaded inserts, base and chemical resistant pre-slit mat
    U-2D™ rack with 96 well plate, 350 µl preloaded inserts, base and chemical resistant pre-slit mat
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 106,00 €
    106,00 €
    1 * 1 ST
       106,00 €
  • MICS95296-500-1P
    U-2D™ rack with 96 well plate, 500 µl preloaded inserts, base and plug-style non-slit silicone sealing mat
    U-2D™ rack with 96 well plate, 500 µl preloaded inserts, base and plug-style non-slit silicone sealing mat
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 134,00 €
    134,00 €
    1 * 1 ST
       134,00 €
  • MICS95296-500-1R
    U-2D™ rack with 96 well plate, 500 µl preloaded inserts, base and chemical resistant non-slit mat
    U-2D™ rack with 96 well plate, 500 µl preloaded inserts, base and chemical resistant non-slit mat
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 140,00 €
    140,00 €
    1 * 1 ST
       140,00 €
  • MICS95296-500-3P
    U-2D™ rack with 96 well plate, 500 µl preloaded inserts, base and plug-style pre-slit silicone sealing mat
    U-2D™ rack with 96 well plate, 500 µl preloaded inserts, base and plug-style pre-slit silicone sealing mat
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 137,00 €
    137,00 €
    1 * 1 ST
       137,00 €
  • MICS95296-500-3R
    U-2D™ rack with 96 well plate, 500 µl preloaded inserts, base and chemical resistant pre-slit mat
    U-2D™ rack with 96 well plate, 500 µl preloaded inserts, base and chemical resistant pre-slit mat
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 144,00 €
    144,00 €
    1 * 1 ST
       144,00 €
  • MICS95296-U2D-1ML
    U-2D™ rack with 96 well glass lined plate, 350 µl preloaded inserts
    U-2D™ rack with 96 well glass lined plate, 350 µl preloaded inserts
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 132,00 €
    132,00 €
    1 * 1 ST
       132,00 €
  • MICS95296-U2D-500
    U-2D™ rack with 96 well glass lined plate, 500 µl preloaded inserts
    U-2D™ rack with 96 well glass lined plate, 500 µl preloaded inserts
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 126,00 €
    126,00 €
    1 * 1 ST
       126,00 €
Racks only
Product Information
  • Availability
    Price
  • MICS95296-350-CS
    U-2D™ rack with 96 well plate, 350 µl preloaded inserts
    U-2D™ rack with 96 well plate, 350 µl preloaded inserts
    Log in for availability
    Log in for availability
    1 * 18 ST
    1 * 18 ST - 1 760,00 €
    1 760,00 €
    1 * 18 ST
       1 760,00 €
  • MICS95296-350-D-CS
    U-2D™ rack with 96 well plate, 350 µl silanised RSA-Pro preloaded inserts
    U-2D™ rack with 96 well plate, 350 µl silanised RSA-Pro preloaded inserts
    Log in for availability
    Log in for availability
    1 * 18 ST
    1 * 18 ST - 2 120,00 €
    2 120,00 €
    1 * 18 ST
       2 120,00 €
  • MICS95296-350-D-PK
    U-2D™ rack with 96 well plate, 350 µl silanised RSA-Pro preloaded inserts
    U-2D™ rack with 96 well plate, 350 µl silanised RSA-Pro preloaded inserts
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 753,00 €
    753,00 €
    1 * 6 ST
       753,00 €
  • MICS95296-350-PK
    U-2D™ rack with 96 well plate, 350 µl preloaded inserts
    U-2D™ rack with 96 well plate, 350 µl preloaded inserts
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 651,00 €
    651,00 €
    1 * 6 ST
       651,00 €
  • MICS95296-500-CS
    U-2D™ rack with 96 well plate, 1 ml preloaded inserts
    U-2D™ rack with 96 well plate, 1 ml preloaded inserts
    Log in for availability
    Log in for availability
    1 * 18 ST
    1 * 18 ST - 1 930,00 €
    1 930,00 €
    1 * 18 ST
       1 930,00 €
  • MICS95296-500-PK
    U-2D™ rack with 96 well plate, 500 µl preloaded inserts
    U-2D™ rack with 96 well plate, 500 µl preloaded inserts
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 671,00 €
    671,00 €
    1 * 6 ST
       671,00 €
Mats
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Plug-style sealing mat for 350 µl U-2D™ plate system, non slit silicone rubber
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 97,80 €
    97,80 €
    1 * 5 ST
       97,80 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Plug-style sealing mat for 350 µl U-2D™ plate system, pre-slit silicone rubber
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 125,00 €
    125,00 €
    1 * 5 ST
       125,00 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Plug-style sealing mat for 350 µl U-2D™ plate system, pre-slit silicone rubber
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 25,20 €
    25,20 €
    1 * 1 ST
       25,20 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Plug-style organic chemical resistant sealing mat for 350 µl U-2D™ plate system, pre-slit
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,10 €
    26,10 €
    1 * 1 ST
       26,10 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Organic chemical resistant sealing mat for 350 µl U-2D™ plate system, pre-slit
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 129,00 €
    129,00 €
    1 * 5 ST
       129,00 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Plug-style sealing mat for 350 µl U-2D™ plate system, non slit silicone rubber
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,70 €
    19,70 €
    1 * 1 ST
       19,70 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Organic chemical resistant sealing mat for 350 µl U-2D™ plate system, non slit
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 112,00 €
    112,00 €
    1 * 5 ST
       112,00 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Plug-style organic chemical resistant sealing mat for 350 µl U-2D™ plate system, non slit
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,50 €
    22,50 €
    1 * 1 ST
       22,50 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Paddles for U-2D™ sealing film
    Paddles for U-2D™ sealing film
    MICS95296-PDL-PK
    Log in for availability
    1 * 5 ST - 17,10 €
Product Information
  • Catalog #
    Availability
    Price
  • Roller for U-2D™ sealing film
    Roller for U-2D™ sealing film
    MICS95296-ROLLER
    Log in for availability
    1 * 1 ST - 65,60 €
Product Information
  • Catalog #
    Availability
    Price
  • U-2D™ plate, 350µL, with rack and base only
    U-2D™ plate, 350µL, with rack and base only
    MICS95296-U2D
    Log in for availability
    1 * 1 ST - 116,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Zone free sealing film for U-2D™ plates
    Zone free sealing film for U-2D™ plates
    MICS95296-ZFF-PK
    Log in for availability
    1 * 10 ST - 54,50 €
Product Information
  • Catalog #
    Availability
    Price
  • Zone free sealing film for U-2D™ plates
    Zone free sealing film for U-2D™ plates
    MICS95296-ZFF-CS
    Log in for availability
    1 * 50 ST - 228,00 €