Order Entry
Belgium
ContactUsLinkComponent
Nitrate/ammonium standard solution
Nitrate/ammonium standard solution
  KRAF27278.2000
 :  
Nitrate/ammonium standard solution
  KRAF27278.2000
 :  

 

  • Concentration:
    200 mg N/l
  • Matrix:
    H₂O
  • Pk:
    500 mL
  • Description:
    Nitrate/ammonium standard solution

 

 

Solution with 100 mg each NH₄-N + NO₃-N/l KNO₃ + (NH₄)₂SO₄ in water for TNb determination.