Autosampler vials, inserts and kits, RSA™
About this item
RSA™ (Reduced Surface Activity) high quality and extremely consistent autosampler vials and caps due to reduced surface activity glass manufacturing and control processes. Excellent for low abundance or low dosage form analytes or samples that are pH sensitive, or basic in nature.
- Low adsorption glass, useful for low abundance basic compounds
- No pH change from the vial when using a high amount of water in samples
- Virtually no surface metals prevents adducts in LCMS
- Completely manufactured robotically, not touched by human hands
- Reliability and conistency ensured through extreme QC testing
Available in standard 2 ml sizes, clear and amber formats, and with limited volume features when sample is limited. Improved results with LCMS, GCMS or UHPLC. All lots are tested for fit, form, cleanliness, adsorption, pH control, and metal content.
: Supplied with certificate if requested. Available in "easy purchase packs".
56 Options Available Below
Product Details & Documents
RSA™ (Reduced Surface Activity) high quality and extremely consistent autosampler vials and caps due to reduced surface activity glass manufacturing and control processes. Excellent for low abundance or low dosage form analytes or samples that are pH sensitive, or basic in nature.
- Low adsorption glass, useful for low abundance basic compounds
- No pH change from the vial when using a high amount of water in samples
- Virtually no surface metals prevents adducts in LCMS
- Completely manufactured robotically, not touched by human hands
- Reliability and conistency ensured through extreme QC testing
Available in standard 2 ml sizes, clear and amber formats, and with limited volume features when sample is limited. Improved results with LCMS, GCMS or UHPLC. All lots are tested for fit, form, cleanliness, adsorption, pH control, and metal content.
: Supplied with certificate if requested. Available in "easy purchase packs".
Product Family Options
RSA™ vials
Product Information
- CapacityØ×HColourCap includedCap typeCap colourSeptumAvailabilityPrice
- MICS9532C-0CV-RSSnap/crimp top vial, fused insert, with write-on patchSnap/crimp top vial, fused insert, with write-on patch0,30 ml12×32 mmClearNoLog in for availability1 * 100 ST257,00 €Specifications:
More
Specifications
Cat. No.MICS9532C-0CV-RS - MICS9532S-0CV-RSScrew top vial, fused insert, with write-on patchScrew top vial, fused insert, with write-on patch0,30 ml12×32 mmClearNoLog in for availability1 * 100 ST265,00 €Specifications:
More
Specifications
Cat. No.MICS9532S-0CV-RS - MICS9532S-WCV-WTRSScrew top vial, wide tip fused insert, with write-on patchScrew top vial, wide tip fused insert, with write-on patch0,30 ml12×32 mmClearNoLog in for availability1 * 100 ST256,00 €Specifications:
More
Specifications
Cat. No.MICS9532S-WCV-WTRS - MICS9512C-0CV-T-RSSnap/crimp top vial, MRQSpecifications:
More
Specifications
Cat. No.MICS9512C-0CV-T-RS - MICS9512S-0CV-T-RSScrew top vial (9-425 threads), MRQScrew top vial (9-425 threads), MRQ1,20 ml12×32 mmClearNoLog in for availability1 * 100 ST262,00 €Specifications:
More
Specifications
Cat. No.MICS9512S-0CV-T-RS - MICS9512C-0CV-RSSnap/crimp top vial, maximum recoverySnap/crimp top vial, maximum recovery1,80 ml12×32 mmClearNoLog in for availability1 * 100 ST288,00 €Specifications:
More
Specifications
Cat. No.MICS9512C-0CV-RS - MICS9512S-0CV-RSScrew top vial (9-425 threads), maximum recoveryScrew top vial (9-425 threads), maximum recovery1,80 ml12×32 mmClearNoLog in for availability1 * 100 ST283,00 €Specifications:
More
Specifications
Cat. No.MICS9512S-0CV-RS - MICS9509C-WAV-RSSnap/crimp top vial, with write-on patchSnap/crimp top vial, with write-on patch2,00 ml12×32 mmAmberNoLog in for availability1 * 100 ST48,10 €Specifications:
More
Specifications
Cat. No.MICS9509C-WAV-RS - MICS9509C-WCV-RSSnap/crimp top vial, with write-on patchSnap/crimp top vial, with write-on patch2,00 ml12×32 mmClearNoLog in for availability1 * 100 ST45,20 €Specifications:
More
Specifications
Cat. No.MICS9509C-WCV-RS - MICS9509S-0CV-RSScrew top vial (9-425 threads)Specifications:
More
Specifications
Cat. No.MICS9509S-0CV-RS - MICS9509S-WAV-RSScrew top vial (9-425 threads), with write-on patchScrew top vial (9-425 threads), with write-on patch2,00 ml12×32 mmAmberNoLog in for availability1 * 100 ST49,30 €Specifications:
More
Specifications
Cat. No.MICS9509S-WAV-RS - MICS9509S-WCV-RSScrew top vial (9-425 threads), with write-on patchScrew top vial (9-425 threads), with write-on patch2,00 ml12×32 mmClearNoLog in for availability1 * 100 ST45,30 €Specifications:
More
Specifications
Cat. No.MICS9509S-WCV-RS
Easy purchase packs
Product Information
- CapacityØ×HColourCap includedCap typeCap colourSeptumAvailabilityPrice
- MICS9532C-1CP-RSSnap top vial kits, fused insertSnap top vial kits, fused insert0,30 ml12×32 mmClearYesSnap cap, AQR™Light blueSilicone/PTFE, fittedLog in for availability1 * 100 ST305,00 €Specifications:
More
Specifications
Cat. No.MICS9532C-1CP-RS - MICS9532C-3CP-RSSnap top vial kits, fused insertSnap top vial kits, fused insert0,30 ml12×32 mmClearYesSnap capBlueSilicone/PTFE, pre-slit, fittedLog in for availability1 * 100 ST314,00 €Specifications:
More
Specifications
Cat. No.MICS9532C-3CP-RS - MICS9532S-1CP-RSScrew top vial kits, wide tip fused insertScrew top vial kits, wide tip fused insert0,30 ml12×32 mmClearYesScrew cap, AQR™Light blueSilicone/PTFE, fittedLog in for availability1 * 100 ST313,00 €Specifications:
More
Specifications
Cat. No.MICS9532S-1CP-RS - MICS9532S-1CPWT-RSScrew top vial kits, fused insertScrew top vial kits, fused insert0,30 ml12×32 mmClearYesScrew cap, AQR™Light blueSilicone/PTFE, fittedLog in for availability1 * 100 ST304,00 €Specifications:
More
Specifications
Cat. No.MICS9532S-1CPWT-RS - MICS9532S-3CP-RSScrew top vial kits, fused insertScrew top vial kits, fused insert0,30 ml12×32 mmClearYesScrew cap, AQR™Light blueSilicone/PTFE, fittedLog in for availability1 * 100 ST317,00 €Specifications:
More
Specifications
Cat. No.MICS9532S-3CP-RS - MICS9532S-3CPWT-RSScrew top vial kits, wide tip fused insertScrew top vial kits, wide tip fused insert0,30 ml12×32 mmClearYesScrew cap, AQR™Light blueSilicone/PTFE, pre-slit, fittedLog in for availability1 * 100 ST336,00 €Specifications:
More
Specifications
Cat. No.MICS9532S-3CPWT-RS - MICS9512S-1MP-RSScrew top vial kits, MRQ maximum recoveryScrew top vial kits, MRQ maximum recovery1,20 ml12×32 mmClearYesScrew cap, AQR™Light blueSilicone/PTFE, fittedLog in for availability1 * 100 ST301,00 €Specifications:
More
Specifications
Cat. No.MICS9512S-1MP-RS - MICS9512C-1MP-RSSnap top vial kits, maximum recovery RSA™ glassSnap top vial kits, maximum recovery RSA™ glass1,50 ml12×32 mmClearYesSnap capBlueSilicone/PTFELog in for availability1 * 100 ST312,00 €Specifications:
More
Specifications
Cat. No.MICS9512C-1MP-RS - MICS9512C-3MP-RSSnap top vial kits, maximum recovery RSA™ glassSnap top vial kits, maximum recovery RSA™ glass1,50 ml12×32 mmClearYesSnap capBlueSilicone/PTFE, pre-slitLog in for availability1 * 100 ST344,00 €Specifications:
More
Specifications
Cat. No.MICS9512C-3MP-RS - MICS9512S-3MP-RSScrew top vial kits, maximum recoveryScrew top vial kits, maximum recovery1,50 ml12×32 mmClearYesScrew cap, AQR™Light blueSilicone/PTFE, pre-slit, fittedLog in for availability1 * 100 ST313,00 €Specifications:
More
Specifications
Cat. No.MICS9512S-3MP-RS - MICS9512C-1CP-RSSnap top vial kits, maximum recovery RSA™ glassSnap top vial kits, maximum recovery RSA™ glass1,80 ml12×32 mmClearYesSnap cap, AQR™Light blueSilicone/PTFE, fittedLog in for availability1 * 100 ST326,00 €Specifications:
More
Specifications
Cat. No.MICS9512C-1CP-RS - MICS9512C-3CP-RSSnap top vial kits, maximum recovery RSA™ glassSnap top vial kits, maximum recovery RSA™ glass1,80 ml12×32 mmClearYesSnap capBlueSilicone/PTFE, pre-slitLog in for availability1 * 100 ST335,00 €Specifications:
More
Specifications
Cat. No.MICS9512C-3CP-RS - MICS9512S-1AP-RSScrew top vial kits, maximum recovery RSA™ glassScrew top vial kits, maximum recovery RSA™ glass1,80 ml12×32 mmAmberYesScrew cap, AQR™Light blueSilicone/PTFE, fittedLog in for availability1 * 100 ST334,00 €Specifications:
More
Specifications
Cat. No.MICS9512S-1AP-RS - MICS9512S-9AP-RSScrew top vial kits, maximum recovery MRQScrew top vial kits, maximum recovery MRQ1,80 ml12×32 mmAmberYesUltra pure PE screw caps (single use)ClearLog in for availability1 * 100 ST326,00 €Specifications:
More
Specifications
Cat. No.MICS9512S-9AP-RS - MICS9512S-9CP-RSScrew top vials kits, with write-on patch, maximum recoveryScrew top vials kits, with write-on patch, maximum recovery1,80 ml12×32 mmClearYesUltra pure PE screw caps (single use)ClearLog in for availability1 * 100 ST331,00 €Specifications:
More
Specifications
Cat. No.MICS9512S-9CP-RS - MICS9512S-1CP-RSScrew top vial kits, maximum recovery RSA™ glassScrew top vial kits, maximum recovery RSA™ glass1,80 ml12×32 mmClearYesScrew cap, AQR™Light blueSilicone/PTFE, fittedLog in for availability1 * 100 ST340,00 €Specifications:
More
Specifications
Cat. No.MICS9512S-1CP-RS - MICS9512S-3AP-RSScrew top vial kits, maximum recoveryScrew top vial kits, maximum recovery1,80 ml12×32 mmAmberYesScrew cap, AQR™Light blueSilicone/PTFE, pre-slit, fittedLog in for availability1 * 100 ST338,00 €Specifications:
More
Specifications
Cat. No.MICS9512S-3AP-RS - MICS9512S-3CP-RSScrew top vial kits, maximum recoveryScrew top vial kits, maximum recovery1,80 ml12×32 mmClearYesScrew cap, AQR™Light blueSilicone/PTFE, pre-slit, fittedLog in for availability1 * 100 ST344,00 €Specifications:
More
Specifications
Cat. No.MICS9512S-3CP-RS - MICS9509C-1WCP-RSSnap top vial kits, with write-on patchSnap top vial kits, with write-on patch2,00 ml12×32 mmClearYesSnap cap, Soft-Guard™BlueSilicone/PTFE, fittedLog in for availability1 * 100 ST87,90 €Specifications:
More
Specifications
Cat. No.MICS9509C-1WCP-RS - MICS9509C-3EWCP-RSSnap top vial kits, with write-on patchSnap top vial kits, with write-on patch2,00 ml12×32 mmClearYesSnap cap, Soft-Guard™BlueSilicone/PTFE, pre-slit, fittedLog in for availability1 * 100 ST84,70 €Specifications:
More
Specifications
Cat. No.MICS9509C-3EWCP-RS - MICS9509S-1EWAP-RSScrew top vial kits, with write-on patchScrew top vial kits, with write-on patch2,00 ml12×32 mmAmberYesScrew cap, Soft-Guard™BlueSilicone/PTFE, bondedLog in for availability1 * 100 ST91,80 €Specifications:
More
Specifications
Cat. No.MICS9509S-1EWAP-RS - MICS9509S-1EWCP-RSScrew top vial kits, with write-on patchScrew top vial kits, with write-on patch2,00 ml12×32 mmClearYesScrew cap, Soft-Guard™BlueSilicone/PTFE, bondedLog in for availability1 * 100 ST87,80 €Specifications:
More
Specifications
Cat. No.MICS9509S-1EWCP-RS - MICS9509S-1WAP-RSScrew top vial kits, with write-on patchScrew top vial kits, with write-on patch2,00 ml12×32 mmAmberYesScrew capBlueSilicone/PTFE, bondedLog in for availability1 * 100 ST90,80 €Specifications:
More
Specifications
Cat. No.MICS9509S-1WAP-RS - MICS9509S-1XWAP-RSSnap top vial kits, with write-on patchSnap top vial kits, with write-on patch2,00 ml12×32 mmClearYesScrew cap, Soft-Guard™BlueSilicone/PTFE, bondedLog in for availability1 * 100 ST94,00 €Specifications:
More
Specifications
Cat. No.MICS9509S-1XWAP-RS - MICS9509S-1XWCP-RSSnap top vial kits, with write-on patchSnap top vial kits, with write-on patch2,00 ml12×32 mmClearYesScrew capBlueSilicone/PTFE, bondedLog in for availability1 * 100 ST90,10 €Specifications:
More
Specifications
Cat. No.MICS9509S-1XWCP-RS - MICS9509S-3EWCP-RSSnap top vial kits, with write-on patchSnap top vial kits, with write-on patch2,00 ml12×32 mmAmberYesScrew cap, Soft-Guard™BlueSilicone/PTFE, pre-slit, bondedLog in for availability1 * 100 ST94,60 €Specifications:
More
Specifications
Cat. No.MICS9509S-3EWCP-RS - MICS9509S-3WCPEARSSnap top vial kits, with write-on patchSnap top vial kits, with write-on patch2,00 ml12×32 mmClearYesScrew cap, Soft-Guard™BlueSilicone/PTFE, pre-slit, bondedLog in for availability1 * 100 ST94,60 €Specifications:
More
Specifications
Cat. No.MICS9509S-3WCPEARS - MICS9509S-3XWAP-RSSnap top vial kits, with write-on patchSnap top vial kits, with write-on patch2,00 ml12×32 mmAmberYesScrew capBlueSilicone/PTFE, pre-slit, bondedLog in for availability1 * 100 ST106,00 €Specifications:
More
Specifications
Cat. No.MICS9509S-3XWAP-RS - MICS9509S-3XWCP-RSSnap top vial kits, with write-on patchSnap top vial kits, with write-on patch2,00 ml12×32 mmClearYesScrew capBlueSilicone/PTFE, pre-slit, bondedLog in for availability1 * 100 ST102,00 €Specifications:
More
Specifications
Cat. No.MICS9509S-3XWCP-RS - MICS9509S-9WAP-RSSnap top vial kits, with write-on patchSnap top vial kits, with write-on patch2,00 ml12×32 mmAmberYesUltra pure PE screw caps (single use)ClearLog in for availability1 * 100 ST84,20 €Specifications:
More
Specifications
Cat. No.MICS9509S-9WAP-RS - MICS9509S-9WCP-RSSnap top vial kits, with write-on patchSnap top vial kits, with write-on patch2,00 ml12×32 mmClearYesUltra pure PE screw caps (single use)ClearLog in for availability1 * 100 ST80,30 €Specifications:
More
Specifications
Cat. No.MICS9509S-9WCP-RS - MICS9509C-3WAP-RSSnap top vial kits, with write-on patchSnap top vial kits, with write-on patch2,00 ml12×32 mmAmberYesSnap cap, AQR™Light blueSilicone/PTFE, pre-slit, fittedLog in for availability1 * 100 ST98,50 €Specifications:
More
Specifications
Cat. No.MICS9509C-3WAP-RS - MICS9509C-3WCP-RSSnap top vial kits, with write-on patchSnap top vial kits, with write-on patch2,00 ml12×32 mmClearYesSnap cap, AQR™Light blueSilicone/PTFE, pre-slit, fittedLog in for availability1 * 100 ST95,60 €Specifications:
More
Specifications
Cat. No.MICS9509C-3WCP-RS - MICS9509C-3XWAP-RSSnap top vial kits, with write-on patchSnap top vial kits, with write-on patch2,00 ml12×32 mmAmberYesSnap cap, AQR™Light blueSilicone/PTFE, pre-slit, fittedLog in for availability1 * 100 ST96,50 €Specifications:
More
Specifications
Cat. No.MICS9509C-3XWAP-RS - MICS9509C-3XWCP-RSSnap top vial kits, with write-on patchSnap top vial kits, with write-on patch2,00 ml12×32 mmClearYesSnap cap, AQR™Light blueSilicone/PTFE, pre-slit, fittedLog in for availability1 * 100 ST93,50 €Specifications:
More
Specifications
Cat. No.MICS9509C-3XWCP-RS - MICS9509S-1CP-RSScrew top vial kits, with write-on patchScrew top vial kits, with write-on patch2,00 ml12×32 mmClearYesScrew cap, AQR™Light blueSilicone/PTFE, fittedLog in for availability1 * 100 ST82,90 €Specifications:
More
Specifications
Cat. No.MICS9509S-1CP-RS - MICS9509S-1WCP-RSAScrew top vial kits, with write-on patchScrew top vial kits, with write-on patch2,00 ml12×32 mmClearYesScrew cap, AQR™Light blueClear silicone/PTFE, fittedLog in for availability1 * 100 ST88,00 €Specifications:
More
Specifications
Cat. No.MICS9509S-1WCP-RSA - MICS9509S-1WCP-RSScrew top vial kits, with write-on patchScrew top vial kits, with write-on patch2,00 ml12×32 mmClearYesScrew cap, AQR™Light blueClear silicone/PTFE, fittedLog in for availability1 * 100 ST81,30 €Specifications:
More
Specifications
Cat. No.MICS9509S-1WCP-RS - MICS9509S-3CP-RSSnap top vial kitsSnap top vial kits2,00 ml12×32 mmClearYesScrew cap, AQR™Light blueSilicone/PTFE, pre-slit, fittedLog in for availability1 * 100 ST86,70 €Specifications:
More
Specifications
Cat. No.MICS9509S-3CP-RS - MICS9509S-3WAP-RSSnap top vial kits, with write-on patchSnap top vial kits, with write-on patch2,00 ml12×32 mmAmberYesScrew cap, AQR™Light blueSilicone/PTFE, pre-slit, fittedLog in for availability1 * 100 ST99,00 €Specifications:
More
Specifications
Cat. No.MICS9509S-3WAP-RS - MICS9509S-3WCP-RSSnap top vial kits, with write-on patch, fused insertSnap top vial kits, with write-on patch, fused insert2,00 ml12×32 mmClearYesScrew cap, AQR™Light blueSilicone/PTFE, pre-slit, fittedLog in for availability1 * 100 ST97,10 €Specifications:
More
Specifications
Cat. No.MICS9509S-3WCP-RS - MICS9509C-1EWCP-RSSnap top vial kits, with write-on patchSnap top vial kits, with write-on patch2,00 ml12×32 mmClearYesSnap capBlueWhite silicone/red PTFE, pre-slitLog in for availability1 * 100 ST80,90 €Specifications:
More
Specifications
Cat. No.MICS9509C-1EWCP-RS - MICS9509C-1XWCP-RSSnap top vial kits, with write-on patchSnap top vial kits, with write-on patch2,00 ml12×32 mmClearYesSnap capBlueWhite silicone/red PTFE, fittedLog in for availability1 * 100 ST87,50 €Specifications:
More
Specifications
Cat. No.MICS9509C-1XWCP-RS - MICS9509S-3EWAP-RSSnap top vial kits, with write-on patchSnap top vial kits, with write-on patch2,00 ml12×32 mmAmberYesSnap capBlueWhite silicone/red PTFE, pre-slit, fittedLog in for availability1 * 100 ST98,50 €Specifications:
More
Specifications
Cat. No.MICS9509S-3EWAP-RS
AQR™ caps
Product Information
- Catalog #SeptumDescriptionMaterialAvailabilityPrice
- Pre-slit, bonded, ultra pure clear silicone/PTFE septaScrew caps (9-425 mm threads), Light blue colourPPLog in for availability1 * 100 ST52,00 €