Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Seals
Sealing Rings
O-rings, FPM
O-rings, FPM
O-rings, FPM
28-9535-45
:
Cytiva
O-rings, FPM
28-9535-45
:
Cytiva
Colour:
Green
Description:
O-Ring, 13,1×1,6 mm, Fluor carbon rubber (FPM)
Seq:
34
Pk:
1