Orders LinkContactUsLinkComponent
Torque wrench
Torque wrench
Torque wrench for BPG columns, Stainless steel 304, 2 - 20 N
Catalog # 18-0251-37
Supplier:   Cytiva
Torque wrench
Catalog # 18-0251-37
Supplier:  Cytiva
Torque wrench
Torque wrench for BPG columns, Stainless steel 304, 2 - 20 N
Price
504,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This torque wrench is suitable for assembly and disassembly of BPG columns.
Specifications
  • Description:
    Torque wrench for BPG columns
  • Material:
    Stainless steel 304
  • Seq:
    30
  • Tightening torque:
    2 - 20 N