Orders LinkContactUsLinkComponent
HPLC columns, Cogent TYPE-C Silica™
Supplier:   MICROSOLV
HPLC columns, Cogent TYPE-C Silica™
Supplier:  MICROSOLV
Certificates
HPLC columns, Cogent TYPE-C Silica™
15 Options Available Below
Product Details & Documents
Cogent TYPE-C Silica™ is HPLC grade silica with a silica hydride surface. Their unique stationary phase will adsorb and desorb solvents commonly used in mobile phases very differently from ordinary silica resulting in excellent benefits to the chromatographer. Excellent for normal phase chromatography and solvent drying is not needed.
  • Totally inorganic
  • Unbonded
  • Carbon free
Since these columns have virtually no silanols remaining (less than 2%), they do not have a strong association with water and other solvents and will not have the expected hydration shell of other silica-based columns.
Excellent choice for normal phase and preparative chromatography because they produce a unique selectivity are extremely stable for long column life and can be used to retain some polar compounds. These columns have been used in aqueous normal phase and reversed phase HPLC as well.
Suitable for normal phase HPLC, no need to concern with the moisture content of the solvents.
Specifications for Product Family
Specifications
  • Catalog No:
  • 554-5959
  • 554-5960
  • 554-5958
  • 554-5964
  • 554-5965
  • 554-5963
  • 554-5969
  • 554-5970
  • MICS40000-25P-3
  • 554-5973
  • 554-6413
  • 554-6416
  • 554-6417
  • 554-6420
  • 554-6421
  • Carbon load:
  • 0%
  • 0%
  • 0%
  • 0%
  • 0%
  • 0%
  • 0%
  • 0%
  • 0%
  • 0%
  • 0%
  • 0%
  • 0%
  • 0%
  • 0%
  • Endcapped:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • pH range:
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • 2 - 7
  • Surface area:
  • 390 m²/g
  • 390 m²/g
  • 390 m²/g
  • 390 m²/g
  • 390 m²/g
  • 390 m²/g
  • 390 m²/g
  • 390 m²/g
  • 390 m²/g
  • 390 m²/g
  • 340 m²/g
  • 340 m²/g
  • 340 m²/g
  • 340 m²/g
  • 340 m²/g
  • Temperature limit:
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • USP code:
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
Specifications
Product Family Options
Analytical columns
Product Information
  • Column Format
    Phase
    Column hardware
    Pore size
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • Analytical
    Unmodified Silica Hydride
    Stainless steel
    100 Å
    4 µm
    1,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 060,00 €
    1 060,00 €
    1 * 1 ST
       1 060,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-5959
    Carbon load
    0%
    Endcapped
    No
    pH range
    2 - 7
    Surface area
    390 m²/g
    Temperature limit
    60 °C
    USP code
    L3
  • Analytical
    Unmodified Silica Hydride
    Stainless steel
    100 Å
    4 µm
    2,10 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 838,00 €
    838,00 €
    1 * 1 ST
       838,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-5960
    Carbon load
    0%
    Endcapped
    No
    pH range
    2 - 7
    Surface area
    390 m²/g
    Temperature limit
    60 °C
    USP code
    L3
  • Analytical
    Unmodified Silica Hydride
    Stainless steel
    100 Å
    4 µm
    4,60 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 838,00 €
    838,00 €
    1 * 1 ST
       838,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-5958
    Carbon load
    0%
    Endcapped
    No
    pH range
    2 - 7
    Surface area
    390 m²/g
    Temperature limit
    60 °C
    USP code
    L3
  • Analytical
    Unmodified Silica Hydride
    Stainless steel
    100 Å
    4 µm
    1,00 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 890,00 €
    1 890,00 €
    1 * 1 ST
       1 890,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-5964
    Carbon load
    0%
    Endcapped
    No
    pH range
    2 - 7
    Surface area
    390 m²/g
    Temperature limit
    60 °C
    USP code
    L3
  • Analytical
    Unmodified Silica Hydride
    Stainless steel
    100 Å
    4 µm
    2,10 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 880,00 €
    880,00 €
    1 * 1 ST
       880,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-5965
    Carbon load
    0%
    Endcapped
    No
    pH range
    2 - 7
    Surface area
    390 m²/g
    Temperature limit
    60 °C
    USP code
    L3
  • Analytical
    Unmodified Silica Hydride
    Stainless steel
    100 Å
    4 µm
    4,60 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 010,00 €
    1 010,00 €
    1 * 1 ST
       1 010,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-5963
    Carbon load
    0%
    Endcapped
    No
    pH range
    2 - 7
    Surface area
    390 m²/g
    Temperature limit
    60 °C
    USP code
    L3
  • Analytical
    Unmodified Silica Hydride
    Stainless steel
    100 Å
    4 µm
    1,00 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 890,00 €
    1 890,00 €
    1 * 1 ST
       1 890,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-5969
    Carbon load
    0%
    Endcapped
    No
    pH range
    2 - 7
    Surface area
    390 m²/g
    Temperature limit
    60 °C
    USP code
    L3
  • Analytical
    Unmodified Silica Hydride
    Stainless steel
    100 Å
    4 µm
    2,10 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 060,00 €
    1 060,00 €
    1 * 1 ST
       1 060,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-5970
    Carbon load
    0%
    Endcapped
    No
    pH range
    2 - 7
    Surface area
    390 m²/g
    Temperature limit
    60 °C
    USP code
    L3
  • Analytical
    Unmodified silica hydride
    Stainless steel
    100 Å
    4 µm
    3,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 140,00 €
    1 140,00 €
    1 * 1 ST
       1 140,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MICS40000-25P-3
    Carbon load
    0%
    Endcapped
    No
    pH range
    2 - 7
    Surface area
    390 m²/g
    Temperature limit
    60 °C
    USP code
    L3
  • Analytical
    Unmodified Silica Hydride
    Stainless steel
    100 Å
    4 µm
    4,60 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 110,00 €
    1 110,00 €
    1 * 1 ST
       1 110,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-5973
    Carbon load
    0%
    Endcapped
    No
    pH range
    2 - 7
    Surface area
    390 m²/g
    Temperature limit
    60 °C
    USP code
    L3
2.ō columns
Product Information
  • Column Format
    Phase
    Column hardware
    Pore size
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • Analytical
    Unmodified Silica Hydride
    Stainless steel
    120 Å
    2,2 µm
    4,60 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 080,00 €
    1 080,00 €
    1 * 1 ST
       1 080,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-6413
    Carbon load
    0%
    Endcapped
    No
    pH range
    2 - 7
    Surface area
    340 m²/g
    Temperature limit
    60 °C
    USP code
    L3
  • Analytical
    Unmodified Silica Hydride
    Stainless steel
    120 Å
    2,2 µm
    1,00 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 510,00 €
    1 510,00 €
    1 * 1 ST
       1 510,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-6416
    Carbon load
    0%
    Endcapped
    No
    pH range
    2 - 7
    Surface area
    340 m²/g
    Temperature limit
    60 °C
    USP code
    L3
  • Analytical
    Unmodified Silica Hydride
    Stainless steel
    120 Å
    2,2 µm
    2,10 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 300,00 €
    1 300,00 €
    1 * 1 ST
       1 300,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-6417
    Carbon load
    0%
    Endcapped
    No
    pH range
    2 - 7
    Surface area
    340 m²/g
    Temperature limit
    60 °C
    USP code
    L3
  • Analytical
    Unmodified Silica Hydride
    Stainless steel
    120 Å
    2,2 µm
    1,00 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 510,00 €
    1 510,00 €
    1 * 1 ST
       1 510,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-6420
    Carbon load
    0%
    Endcapped
    No
    pH range
    2 - 7
    Surface area
    340 m²/g
    Temperature limit
    60 °C
    USP code
    L3
  • Analytical
    Unmodified Silica Hydride
    Stainless steel
    120 Å
    2,2 µm
    2,10 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 500,00 €
    1 500,00 €
    1 * 1 ST
       1 500,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    554-6421
    Carbon load
    0%
    Endcapped
    No
    pH range
    2 - 7
    Surface area
    340 m²/g
    Temperature limit
    60 °C
    USP code
    L3