Syringe filters, colour coded
Supplier: MICROSOLV
About this item
Colour coded low volume syringe filter for HPLC, CE or dissolution testing.
- Hydrophilic membrane
- Houding made from virgin medical grade PP
- Retention volume of 90 µl
3 Options Available Below
Product Details & Documents
Colour coded low volume syringe filter for HPLC, CE or dissolution testing.
- Hydrophilic membrane
- Houding made from virgin medical grade PP
- Retention volume of 90 µl
Specifications for Product Family
Specifications
- Catalog No:
- MICS58045-N25-C
- MICS58045-R25-C
- MICS58045-P25-C
- Max. operating pressure:
- 7,6 bar
- 7,6 bar
- 7,6 bar
- Max. temperature:
- 100 °C
- 100 °C
- 100 °C
Specifications
Product Family Options
Product Information
- ØPore sizeMembraneHousing materialColourAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.MICS58045-N25-CMax. operating pressure7,6 barMax. temperature100 °C - Specifications:
More
Specifications
Cat. No.MICS58045-R25-CMax. operating pressure7,6 barMax. temperature100 °C - Specifications:
More
Specifications
Cat. No.MICS58045-P25-CMax. operating pressure7,6 barMax. temperature100 °C