Spray gun
Spray gun, length 278 mm, 28 mm
Catalog # RIXI8-3201-28278BL
Supplier: BERLIN PACKAGING RIXIUS GMBH
Jump to:
Product Details & Documents
Spray gun made up of polypropylene.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
- Thread:28 mm
- Description:Spray gun, length 278 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



