Order Entry
Belgium
ContactUsLinkComponent
tert-Butyl methyl ether ≥99.0% (by GC, corrected for water content), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Flammable
Irritant
Health
tert-Butyl methyl ether ≥99.0% (by GC, corrected for water content), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # 5398-25
CAS Number:  
undefined
Flammable
Irritant
Health
tert-Butyl methyl ether ≥99.0% (by GC, corrected for water content), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # 5398-25
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    ≥99.0%
  • ACS - Assay (by GC, corrected for water):
    ≥99.0 %
  • ACS - Color (APHA):
    ≤10
  • ACS - Peroxide (as H₂O₂):
    ≤1 ppm
  • ACS - Residue after Evaporation:
    ≤2 ppm
  • ACS - Water (H₂O)(by Karl Fischer titrn):
    ≤0.05 %
  • Optical Absorbance (1-cm path vs water) - 210 nm:
    ≤1.00
  • Optical Absorbance (1-cm path vs water) - 225 nm:
    ≤0.50
  • Optical Absorbance (1-cm path vs water) - 250 nm:
    ≤0.10
  • Optical Absorbance (1-cm path vs water) - 300 nm:
    ≤0.005
  • Optical Absorbance (1-cm path vs water) - 350 nm:
    ≤0.005
  • Optical Absorbance (1-cm path vs water) - 400 nm:
    ≤0.005

Specification Test Results

About this item

 :
 :88.15 g/mol
Boiling Pt:55.2 °C
Density:0.74
Flash Pt:–30 °C
Storage Temperature:Ambient
UN:2398
ADR:3,
 :GHS02
 :GHS07
 :GHS08
CAS Number:1634-04-4
Formula:(CH₃)₃COCH₃
MDL Number:MFCD00008812