Multi-element standard solution, 7 elements
Catalog # KRAF04379.2000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Matrix:Nitric acid dil.
- Pk:500 mL
- Description:Multi-elements standard solution for 7 elements
Specifications
About this item
This standard solution is intended for analysis of sludge.
Multi element standards for Zn, Cr, Cu, Pb, Ni, Hg and Cd diluted in nitric acid.