ContactUsLinkComponent
Silver capsule for oxygen analysis kit
Silver capsule for oxygen analysis kit
Catalog # PERCN2411364
Supplier:  PerkinElmer
Silver capsule for oxygen analysis kit
Catalog # PERCN2411364
Supplier:  PerkinElmer
Specifications
  • Description:
    Silver capsules for oxygen analysis kit
  • Pk:
    96
Specifications
About this item
Silver capsule used for analysing organic substances. For oxygen content either silver or standard tin capsules may be used for analysing organic substances for oxygen content.