Multi-element quality control standards for ICP
Supplier: CPACHEM
Jump to:
About this item
ICP QC standards used for Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Cs, Co, Cr, Cu, Fe, In, K, Li, Mg, Mn, Mo, Na, Ni, Nb, Pb, Rb, Sb, Se, Sr, Ti, Tl, V, U, Zn. 1 mg/l for 33 elememts.
2 Options Available Below
Product Details & Documents
ICP QC standards used for Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Cs, Co, Cr, Cu, Fe, In, K, Li, Mg, Mn, Mo, Na, Ni, Nb, Pb, Rb, Sb, Se, Sr, Ti, Tl, V, U, Zn. 1 mg/l for 33 elememts.
Certificates
Search by Lot NumberProduct Family Options
Product Information
- MatrixPkAvailabilityPrice
- ACSDM8A96.1.5N.L1Multi-element quality control standards for 33 elementsMulti-element quality control standards for 33 elements5% HNO₃100 mLLog in for availability1 * 100 mL140,00 €
- ACSDM8A96.1.5N.L5Multi-element quality control standards for 33 elementsMulti-element quality control standards for 33 elements5% HNO₃500 mLLog in for availability1 * 500 mL322,00 €