Order Entry
Belgium
ContactUsLinkComponent
GC columns, glass beads
GC columns, glass beads
  HEWLG3591-81178
 :  Agilent
GC columns, glass beads
  HEWLG3591-81178
 :  Agilent

 

  • Column Format:
    Capillary
  • Ext.Ø:
    3,18 mm
  • Ø int.:
    2,00 mm
  • Mesh size:
    60/80
  • Length:
    1 m
  • Packing material:
    Glass beads
  • Pk:
    1

 

 

Packed GC columns are designed and manufactured to offer excellent and reproducible performance for all sample types associated with packed column separations, most important in the hydrocarbon processing industry.

  • Highly efficient and rigorous packing technology assures column-to-column reproducibility and ultimate efficiency
  • UltiMetal treated stainless steel tubing allows for improved inertness and peak shape performance

High-efficiency packing ensures excellent reproducibility in pressure drop, flow characteristics, and separation performance resulting in less system readjustment after column installation.