



Phthalic anhydride 160 g/l in pyridine, BAKER ANALYZED®, J.T.Baker®
Catalog # 7526.2500
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass container
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Color (APHA):Max. 100
Specification Test Results
About this item
:
:148.12 g/mol
Flash Pt:17 °C
UN:2924
ADR:3 (8),II
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:85-44-9
Formula:C₈H₄O₃
MDL Number:MFCD00005918