TraceCERT®, Supelco® Cyanide IC Standard
Catalogus # SUPL90157-100ML
Leverancier: Merck
CAS-nummer:
Specificaties
- Concentration:1000 mg/L
- Matrix:H₂O
- Pk:100 mL
- Description:Cyanide standard solution
Specificaties
Over dit artikel
1000 mg/L cyanide in water, prepared with high purity K₂[Zn(CN)₄] and water.
Cyanide exists mainly in the complex form [Zn(CN)₄]²⁻ in the water (basic) solution.
Certificeringen: Certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). NIST or BAM Traceable.