Order Entry
Belgium
ContactUsLinkComponent
Potassium hydrogen phthalate 99.95-100.05% (by titrimetric analysis on dry substance), white crystalline powder ACS secondary standard for volumetric titrations
Potassium hydrogen phthalate 99.95-100.05% (by titrimetric analysis on dry substance), white crystalline powder ACS secondary standard for volumetric titrations
Catalog # APPC241481.1608
Supplier:  PanReac AppliChem
CAS Number:  
undefined
Potassium hydrogen phthalate 99.95-100.05% (by titrimetric analysis on dry substance), white crystalline powder ACS secondary standard for volumetric titrations
Catalog # APPC241481.1608
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401
  • Pk Info:
    1×100 g

Specifications

About this item

 :
 :204.22 g/mol
Melting Pt:295…300 °C
Storage Temperature:Ambient
CAS Number:877-24-7
Formula:HOOCC₆H₄COOK
MDL Number:MFCD00013070