
Sodium hydroxide 0.1 N, volumetric solution, traceable to NIST Standard Reference Material (SRM)
Supplier: PanReac AppliChem
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Density:1.004 kg/l
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
:GHS05
Specification Test Results
- Titer (20 °C):0.999 - 1.001
Specification Test Results
Frequently Bought Together