Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes, Unisette™, M405

Tissue processing/embedding cassettes, Unisette™, M405

Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Unisette™, M405
Supplier:  Simport Scientific
Certificates
Tissue processing/embedding cassettes, Unisette™, M405
Tissue processing/embedding cassettes, Unisette™, M405

22 Options Available Below

Product Details & Documents

These cassettes are moulded from a special high density polymer and keep specimens safely submerged in liquid.

  • Resistant to the chemical action of most histological solvents
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • One-piece integral lid eliminates the need for separate steel lids
  • Snap latch and hinge-lock design prevent early separation of base and lid

Lids can be opened and closed as often as necessary and they always relock securely without danger of specimen loss.

Ordering information: Available in non-cytotoxic, non-metallic colors.

Packaging: Available in 25 stacks of 40 cassettes or 3 dispenser boxes of 500 cassettes.

Product Family Options

In dispenser boxes

Product Information

  • Colour
    Availability
    Price
  • White
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 231,00 €
    231,00 €
    1 * 1.500 ST
       231,00 €

  • Pink
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 231,00 €
    231,00 €
    1 * 1.500 ST
       231,00 €

  • Green
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 231,00 €
    231,00 €
    1 * 1.500 ST
       231,00 €

  • Yellow
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 231,00 €
    231,00 €
    1 * 1.500 ST
       231,00 €

  • Blue
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 231,00 €
    231,00 €
    1 * 1.500 ST
       231,00 €

  • Pale orange
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 231,00 €
    231,00 €
    1 * 1.500 ST
       231,00 €

  • Cream
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 231,00 €
    231,00 €
    1 * 1.500 ST
       231,00 €

  • Grey
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 231,00 €
    231,00 €
    1 * 1.500 ST
       231,00 €

  • Purple
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 231,00 €
    231,00 €
    1 * 1.500 ST
       231,00 €

  • Orange
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 231,00 €
    231,00 €
    1 * 1.500 ST
       231,00 €

  • Turquoise
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 231,00 €
    231,00 €
    1 * 1.500 ST
       231,00 €

In stacks

Product Information

  • Colour
    Availability
    Price
  • White
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 233,00 €
    233,00 €
    1 * 1.000 ST
       233,00 €

  • Pink
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 233,00 €
    233,00 €
    1 * 1.000 ST
       233,00 €

  • Green
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 233,00 €
    233,00 €
    1 * 1.000 ST
       233,00 €

  • Yellow
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 233,00 €
    233,00 €
    1 * 1.000 ST
       233,00 €

  • Blue
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 233,00 €
    233,00 €
    1 * 1.000 ST
       233,00 €

  • Pale orange
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 233,00 €
    233,00 €
    1 * 1.000 ST
       233,00 €

  • Cream
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 233,00 €
    233,00 €
    1 * 1.000 ST
       233,00 €

  • Grey
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 233,00 €
    233,00 €
    1 * 1.000 ST
       233,00 €

  • Purple
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 233,00 €
    233,00 €
    1 * 1.000 ST
       233,00 €

  • Orange
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 233,00 €
    233,00 €
    1 * 1.000 ST
       233,00 €

  • Turquoise
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 233,00 €
    233,00 €
    1 * 1.000 ST
       233,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.