Mouse Recombinant MIP-2 (from E. coli)
About this item
Insulin-like growth factor II (IGF-II) is an important fetal growth hormone made by theca cells during gestation. IGF-II engages the IGF-I receptor (IGF1R) to mediate embryonic growth. IGF-II also binds the sink IGF-II receptor (IGF2R) leading to IGF-II degradation.
- High quality
- Low endotoxin
- Affordable
Macrophage Inflammatory Protein 2 (MIP-2), also known as CXCL2, is a small cytokine that is secreted by monocytes and neutrophils at sites of inflammation. MIP-2 functions through the chemokine receptor CXCR2 to act as a chemotactic agent for leukocytes and hematopoietic cells.
Delivery information: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Caution: For research use only.
5 Options Available Below
Product Details & Documents
Insulin-like growth factor II (IGF-II) is an important fetal growth hormone made by theca cells during gestation. IGF-II engages the IGF-I receptor (IGF1R) to mediate embryonic growth. IGF-II also binds the sink IGF-II receptor (IGF2R) leading to IGF-II degradation.
- High quality
- Low endotoxin
- Affordable
Macrophage Inflammatory Protein 2 (MIP-2), also known as CXCL2, is a small cytokine that is secreted by monocytes and neutrophils at sites of inflammation. MIP-2 functions through the chemokine receptor CXCR2 to act as a chemotactic agent for leukocytes and hematopoietic cells.
Delivery information: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Caution: For research use only.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- SHBT200-80-5UG
- SHBT200-80-20UG
- SHBT200-80-100UG
- SHBT200-80-500UG
- SHBT200-80-1MG
- Protein/peptide name:
- MIP-2
- MIP-2
- MIP-2
- MIP-2
- MIP-2
- Protein synonyms:
- CXCL2
- CXCL2
- CXCL2
- CXCL2
- CXCL2
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- P10889
- P10889
- P10889
- P10889
- P10889
- Sequence:
- AVVASELRCQ CLKTLPRVDF KNIQSLSVTP PGPHCAQTEV IATLKGGQKV CLDPEAPLVQ KIIQKILNKG KAN
- AVVASELRCQ CLKTLPRVDF KNIQSLSVTP PGPHCAQTEV IATLKGGQKV CLDPEAPLVQ KIIQKILNKG KAN
- AVVASELRCQ CLKTLPRVDF KNIQSLSVTP PGPHCAQTEV IATLKGGQKV CLDPEAPLVQ KIIQKILNKG KAN
- AVVASELRCQ CLKTLPRVDF KNIQSLSVTP PGPHCAQTEV IATLKGGQKV CLDPEAPLVQ KIIQKILNKG KAN
- AVVASELRCQ CLKTLPRVDF KNIQSLSVTP PGPHCAQTEV IATLKGGQKV CLDPEAPLVQ KIIQKILNKG KAN
- Protease-free:
- N
- N
- N
- N
- N
- Animal-free:
- N
- N
- N
- N
- N
- Nuclease-free:
- N
- N
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- N
- Endotoxin level:
- Low
- Low
- Low
- Low
- Low
- Endotoxin content:
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- UniProtKB:
- P10889
- P10889
- P10889
- P10889
- P10889
- Biological activity:
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Purity:
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- Grade:
- RUO
- RUO
- RUO
- RUO
- RUO
- Formulation:
- 10 mM sodium phosphate, pH 7,5
- 10 mM sodium phosphate, pH 7,5
- 10 mM sodium phosphate, pH 7,5
- 10 mM sodium phosphate, pH 7,5
- 10 mM sodium phosphate, pH 7,5
- Molecular weight:
- 7,9 kDa
- 7,9 kDa
- 7,9 kDa
- 7,9 kDa
- 7,9 kDa
- Reconstitution instructions:
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Storage conditions:
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
- RT
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.SHBT200-80-5UGProtein/peptide nameMIP-2Protein synonymsCXCL2Protein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDP10889SequenceAVVASELRCQ CLKTLPRVDF KNIQSLSVTP PGPHCAQTEV IATLKGGQKV CLDPEAPLVQ KIIQKILNKG KANProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1UniProtKBP10889Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation10 mM sodium phosphate, pH 7,5Molecular weight7,9 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRT - Specifications:
More
Specifications
Cat. No.SHBT200-80-20UGProtein/peptide nameMIP-2Protein synonymsCXCL2Protein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDP10889SequenceAVVASELRCQ CLKTLPRVDF KNIQSLSVTP PGPHCAQTEV IATLKGGQKV CLDPEAPLVQ KIIQKILNKG KANProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1UniProtKBP10889Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation10 mM sodium phosphate, pH 7,5Molecular weight7,9 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRT - Specifications:
More
Specifications
Cat. No.SHBT200-80-100UGProtein/peptide nameMIP-2Protein synonymsCXCL2Protein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDP10889SequenceAVVASELRCQ CLKTLPRVDF KNIQSLSVTP PGPHCAQTEV IATLKGGQKV CLDPEAPLVQ KIIQKILNKG KANProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1UniProtKBP10889Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation10 mM sodium phosphate, pH 7,5Molecular weight7,9 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRT - Specifications:
More
Specifications
Cat. No.SHBT200-80-500UGProtein/peptide nameMIP-2Protein synonymsCXCL2Protein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDP10889SequenceAVVASELRCQ CLKTLPRVDF KNIQSLSVTP PGPHCAQTEV IATLKGGQKV CLDPEAPLVQ KIIQKILNKG KANProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1UniProtKBP10889Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation10 mM sodium phosphate, pH 7,5Molecular weight7,9 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRT - Specifications:
More
Specifications
Cat. No.SHBT200-80-1MGProtein/peptide nameMIP-2Protein synonymsCXCL2Protein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDP10889SequenceAVVASELRCQ CLKTLPRVDF KNIQSLSVTP PGPHCAQTEV IATLKGGQKV CLDPEAPLVQ KIIQKILNKG KANProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1UniProtKBP10889Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation10 mM sodium phosphate, pH 7,5Molecular weight7,9 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRT
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



