Order Entry
Belgium
ContactUsLinkComponent
VWR® PerfectBlue Mini L, Gel System
VWR® PerfectBlue Mini L, Gel System
Catalog # 700-0777
Supplier:  VWR Collection
undefined
VWR® PerfectBlue Mini L, Gel System
Catalog # 700-0777
Supplier:  VWR Collection

Specifications

  • Description:
    PerfectBlue gel system Mini L
  • Seq:
    01
  • Model:
    Mini L
  • Gel volume:
    ~85 ml
  • Buffer volume:
    ~800 ml
  • Run time:
    ≥60 h
  • Gel size W×L:
    120×140 mm
  • Sample capacity:
    8 – 100
  • Unit W×L×H:
    170×250×95 mm
  • Pk:
    1

Specifications

About this item

Capable of running 12×14 cm gels, the largest of the 'Mini' range still offers the convenience of direct gel casting in the tray, in an easy to manage product size for reliable, high quality electrophoresis. This system can be supplied with a choice of gel trays suitable for 2, 4 or 12 combs, and an expansive range of standard and microtitre combs, making this versatile system appropriate for both routine and small separation electrophoresis.

*Based on an agarose gel of approximately 5 mm thickness

Ordering information: System is supplied with gasketed UV transmissible (UVT) gel tray (L4) with 4 comb positions and 2 combs (12 and 20 teeth, 1,5 mm thickness) as standard.

Combs

Product Information