Order Entry
Belgium
ContactUsLinkComponent
 
VWR® Midi S PerfectBlue, Gel system
  700-0814
undefined
VWR® Midi S PerfectBlue, Gel system
  700-0814

 

  • Description:
    PerfectBlue gel system Midi S
  • Model:
    Midi S
  • Gel volume:
    ~120 ml
  • Buffer volume:
    ~1000 ml
  • Run time:
    ≥45 h
  • Gel size W×L:
    150×150 mm
  • Sample capacity:
    10 – 160
  • Unit W×L×H:
    215×275×100 mm
  • Pk:
    1

 

 

Capable of rapidly running up to 160 samples, the Midi S is a compact, but highly efficient, electrophoresis system. Versatility is built-in, with six different comb positions and a choice of 10 different comb types and thicknesses available. For rapid handling, special microtitre combs enable rapid loading with a multichannel pipette, while the effective rubber sealed 'end gates' allow gel pouring right in the tray. With its sturdy design and straight electrodes, the results are clear, crisp and quick, free from 'smiling' bands.

*Based on an agarose gel of approximately 5 mm thickness

 : Supplied with 4 combs (1,5 mm thickness, 2×17 and 2×34 teeth), lid, gel tray, tank, cables and 'end gates' for gel pouring.

Combs