Order Entry
Belgium
ContactUsLinkComponent
 
Human recombinant FGF-21 (from E. coli)
 
undefined
Human recombinant FGF-21 (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
    Bioactivity assay: NIH-3T3 proliferation with β-klotho and heparin, ED50 ≤500 ng/ml, Bioactivity ≥2,0 ×10³ unit/mg
  • Gene ID:
    Q9NSA1
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    FGFL
  • Protein/peptide name:
    FGF-21
  • Purity:
    ≥95%
  • Molecular weight:
    19,5 kDa
  • Sequence:
    MHPIPDSSPL LQFGGQVRQR YLYTDDAQQT EAHLEIREDG TVGGAADQSP ESLLQLKALK PGVIQILGVK TSRFLCQRPD GALYGSLHFD PEACSFRELL LEDGYNVYQS EAHGLPLHLP GNKSPHRDPA PRGPARFLPL PGLPPALPEP PGILAPQPPD VGSSDPLSMV GPSQGRSPSY AS
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, 100 mM sodium chloride, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

 

Frequently Bought Together