

Hexamethylenetetramine 98.5% contains an anticaking agent
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-97-0
:
Formula:C₆H₁₂N₄
MDL Number:MFCD00006895
Merck Index:13,05994
Melting Pt:260 °C (subl.)
Density:1.33 g/cm³
Flash Pt:250 °C
Storage Temperature:Ambient
EINECS:202-905-8
ADR:4.1,III
:140.19 g/mol
UN:1328
:GHS07
:GHS02
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Water:≤0.5 % (K.F.)
- Ash:0.5 % to 3.0 % (anticaking agent)
- Titration excess HCl:≥98.0 %
Specification Test Results
Frequently Bought Together