


HMDS (1,1,1,3,3,3-hexamethyldisilazane)
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:999-97-3
:
Formula:(CH₃)₃SiNHSi(CH₃)₃
MDL Number:MFCD00008259
:161.39 g/mol
Merck Index:13,04708
Boiling Pt:126 °C (1013 hPa)
Melting Pt:–82 °C
Density:0.7742 g/cm³ (20 °C)
Flash Pt:6 °C
Storage Temperature:Ambient
EINECS:213-668-5
UN:2924
ADR:3,II
REACH:01-2119438176-38
:GHS02
:GHS06
:GHS05
Frequently Bought Together