Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Pyridine, extra pure

Flammable
Irritant
Supplier:   Bernd Kraft
Pyridine, extra pure
Supplier:  Bernd Kraft
Certificates
Pyridine, extra pure
Pyridine, extra pure

6 Options Available Below

Product Details & Documents
Synonyms:  Azabenzene, Azine
  • CAS Number:110-86-1
  • Formula:C₅H₅N
  • MDL Number:MFCD00011732
  • Molecular Weight:79.1 g/mol
  • Merck Index:13,08060
  • Boiling Pt:115 °C (1013 hPa)
  • Melting Pt:–44 °C
  • Density:0.983 g/cm³ (20 °C)
  • Flash Pt:17 °C
  • Storage Temperature:Ambient
  • EINECS:203-809-9
  • UN:1282
  • ADR:3,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 46,40 €
    46,40 €
    1 * 250 mL
       46,40 €

  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 75,20 €
    75,20 €
    1 * 500 mL
       75,20 €

  • Glass bottle
    2.500 mL
    Log in for availability
    Log in for availability
    1 * 2.500 mL
    1 * 2.500 mL - 257,00 €
    257,00 €
    1 * 2.500 mL
       257,00 €

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 124,00 €
    124,00 €
    1 * 1 L
       124,00 €

  • 5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 419,00 €
    419,00 €
    1 * 5 L
       419,00 €

  • 25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L - 1 220,00 €
    1 220,00 €
    1 * 25 L
       1 220,00 €

This product has been blocked by your organization. Please contact your purchasing department for more information