Order Entry
Belgium
ContactUsLinkComponent
Mouse Recombinant EXODUS-2/CCL21 (from E. coli)
Mouse Recombinant EXODUS-2/CCL21 (from E. coli)
Certificates
undefined
Mouse Recombinant EXODUS-2/CCL21 (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    P84444
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    CCL21|6Ckine
  • UniProtKB:
    P84444
  • Protein/peptide name:
    Exodus-2/CCL21
  • Purity:
    ≥95%
  • Molecular weight:
    12 kDa
  • Sequence:
    SDGGGQDCCL KYSQKKIPYS IVRGYRKQEP SLGCPIPAIL FSPRKHSKPE LCANPEEGWV QNLMRRLDQP PAPGKQSPGC RKNRGTSKSG KKGKGSKGCK RTEQTQPSRG
  • Endotoxin level:
    Low
  • Formulation:
    0,1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together