Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Nicotinamide

Glass bottle for solids, 100 mg
Catalog # PROOICRS0357
Irritant
Nicotinamide
Catalog # PROOICRS0357
CAS Number:  98-92-0
Nicotinamide

Glass bottle for solids, 100 mg

CAS Number:  98-92-0
Price
367,00 €
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3
  • CAS Number:98-92-0
  • Formula:C₆H₆N₂O
  • MDL Number:MFCD00006395
  • Molecular Weight:122.13 g/mol
  • Merck Index:12,06574
  • Boiling Pt:334 °C (1013 hPa)
  • Melting Pt:127…128 °C
  • Density:1.4463 g/cm³ (20 °C)
  • Flash Pt:182 °C
  • Storage Temperature:Ambient
  • EINECS:202-713-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.