Nicotinamide 99%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3
- CAS Number:98-92-0
- Formula:C₆H₆N₂O
- MDL Number:MFCD00006395
- Molecular Weight:122.13 g/mol
- Merck Index:12,06574
- Boiling Pt:150…160 °C (760 mmHg)
- Melting Pt:128…131 °C
- Density:1,4463 g/cm³ (20 °C)
- Flash Pt:>150 °C
- Storage Temperature:Ambient
- EINECS:202-713-4
3 Options Available Below
Product Details & Documents
Synonyms: 3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3
- CAS Number:98-92-0
- Formula:C₆H₆N₂O
- MDL Number:MFCD00006395
- Molecular Weight:122.13 g/mol
- Merck Index:12,06574
- Boiling Pt:150…160 °C (760 mmHg)
- Melting Pt:128…131 °C
- Density:1,4463 g/cm³ (20 °C)
- Flash Pt:>150 °C
- Storage Temperature:Ambient
- EINECS:202-713-4
Documents
ACRO12827
SDS
ACRO128275000_TDS_EN
Product specification
Specifications for Product Family
Specification Test Results
- Appearance:White to almost white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:128 to 131 °C
- Loss on drying:≤0.5 % (4 hrs) (silica gel)
- Sulfated ash:≤0.1 %
- Readily carbonizable substance:Passes test
- pH:6.0 to 7.5 (5 % at 20 °C)
- Titration with HClO4:≥98.5 % (on dried substance)
- Heavy metals (as Pb):≤30 ppm
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



