Order Entry
Belgium
Orders LinkContactUsLinkComponent
Chromium(III) picolinate

Chromium(III) picolinate

Catalog #
Irritant
Chromium(III) picolinate
Catalog #
Supplier:  
CAS Number:  14639-25-9
Chromium(III) picolinate
CAS Number:  14639-25-9
Product Details & Documents

Picolinic Acid Chromium Salt (CrPic) is a supplemental or alternative medication for type 2 diabetes. Experimental evidence shows the effect of CrPic on glucose uptake through the activation of p38 MAPK. Chromium is thought to potentiate the action of insulin, leading to increased insulin sensitivity in type 2 diabetes.

Picolinic Acid Chromium Salt, a dietay supplement, may be used to study its potential as a modulator of glucose uptake and the activity of insulin. It is used to study its effects on nuclear factor-κ B (NF-κB) and nuclear factor-E2-related factor-2 (Nrf2) pathways in animals.

Very slightly soluble in water (300 mg/litre) and chloroform (900 mg/litre).

Synonyms:  Picolinic acid chromium salt, Chromium picolinate
  • CAS Number:14639-25-9
  • Formula:C₁₈H₁₂CrN₃O₆
  • MDL Number:MFCD00068715
  • Merck Index:12,07557
  • Boiling Pt:293 °C (1013 hPa)
  • Melting Pt:134…137 °C
  • Density:1.49 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:202-719-7