Order Entry
Belgium
ContactUsLinkComponent
Human Recombinant NAP-2 (from E. coli)
Human Recombinant NAP-2 (from E. coli)
New Product
Certificates
undefined
Human Recombinant NAP-2 (from E. coli)
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    P02775
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    Platelet basic protein|MDGF|LDGF|PBP|CXCL7, Neutrophil activating peptide 2|PPBP
  • Protein/peptide name:
    NAP-2
  • Purity:
    ≥95%
  • Molecular weight:
    7,6 kDa
  • Sequence:
    AELRCMCIKT TSGIHPKNIQ SLEVIGKGTH CNQVEVIATL KDGRKICLDP DAPRIKKIVQ KKLAGDESAD
  • Endotoxin level:
    Low
  • Formulation:
    10 mM Sodium phosphate, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together