Order Entry
Belgium
Orders LinkContactUsLinkComponent
Caffeine

Caffeine

Glass bottle for solids, 100 mg
N° de catalogue PROOICRS0234
Irritant
Fournisseur:   LGC Standards PROMOCHEM
Caffeine
N° de catalogue PROOICRS0234
Numéro CAS:  58-08-2
Caffeine
Caffeine

Glass bottle for solids, 100 mg

Numéro CAS:  58-08-2
Prix
367,00 €
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione, 7-Methyltheobromine, 1,3,7-Trimethylxanthine, Methyltheobromine, 1,3,7-Trimethyl-2,6-dioxopurine, 2,6-Dihydroxy-1,3,7-trimethylpurine
  • CAS Number:58-08-2
  • Formula:C₈H₁₀N₄O₂
  • MDL Number:MFCD00005758
  • Molecular Weight:194.19 g/mol
  • Merck Index:13,01636
  • Melting Pt:235…239 °C
  • Density:1.23 g/cm³ (19 °C)
  • Storage Temperature:Ambient
  • EINECS:200-362-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.