Order Entry
Belgium
Orders LinkContactUsLinkComponent
Caffeine

Caffeine

1
Catalog # NENLNEC-412/050UC
Irritant
Marketsource
Supplier:   NEN LIFE SCIENCE
Caffeine
Catalog # NENLNEC-412/050UC
Supplier:  NEN LIFE SCIENCE
Marketsource
CAS Number:  58-08-2
Caffeine
CAS Number:  58-08-2
Price
Inquire For Price
Log in for availability
1 * 1 Un.
Product Details & Documents
Synonyms:  3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione, 7-Methyltheobromine, 1,3,7-Trimethylxanthine, Methyltheobromine, 1,3,7-Trimethyl-2,6-dioxopurine, 2,6-Dihydroxy-1,3,7-trimethylpurine
  • CAS Number:58-08-2
  • Formula:C₈H₁₀N₄O₂
  • MDL Number:MFCD00005758
  • Molecular Weight:194.19 g/mol
  • Merck Index:13,01636
  • Melting Pt:235…239 °C
  • Density:1.23 g/cm³ (19 °C)
  • Storage Temperature:Ambient
  • EINECS:200-362-1
Specifications

Specifications

  • Pk:
    1

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.