Caffeine ≥99%
About this item
Purity: 99% CAS No.: 58-08-2 Molecular Formula: C8H10N4O2 Formula Weight: 194.19 Physical Form: Powder Density: 1.230 Appearance: White Melting Point: 234-236° Solubility: Soluble in pyrrole, THF-H2O mixture, ethyl acetate Beilstein Reference: 17705 Merck Reference: 14,1636 UN No.: 1544 MDL No.: MFCD00005758
Has inhibitory action against lung and colon tumorigenses, and virus induction
- CAS Number:58-08-2
- Formula:C₈H₁₀N₄O₂
- MDL Number:MFCD00005758
- Molecular Weight:194.19 g/mol
- Merck Index:14,1636
- Melting Pt:234…236 °C
- Density:1.230
- Storage Temperature:Ambient
- EINECS:200-362-1
- UN:1544
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 58-08-2 Molecular Formula: C8H10N4O2 Formula Weight: 194.19 Physical Form: Powder Density: 1.230 Appearance: White Melting Point: 234-236° Solubility: Soluble in pyrrole, THF-H2O mixture, ethyl acetate Beilstein Reference: 17705 Merck Reference: 14,1636 UN No.: 1544 MDL No.: MFCD00005758
Has inhibitory action against lung and colon tumorigenses, and virus induction
- CAS Number:58-08-2
- Formula:C₈H₁₀N₄O₂
- MDL Number:MFCD00005758
- Molecular Weight:194.19 g/mol
- Merck Index:14,1636
- Melting Pt:234…236 °C
- Density:1.230
- Storage Temperature:Ambient
- EINECS:200-362-1
- UN:1544
- ADR:6.1,III
Documents
Certificates
Search by Lot NumberALFAA10431.0B_TDS_EN
Product specification
Organic Building Blocks and Intermediates



