Order Entry
Belgium
ContactUsLinkComponent
3-Isobutyl-1-methylxanthine
Irritant
3-Isobutyl-1-methylxanthine
Catalog # SERA26445.02
Supplier:  Serva
CAS Number:  
Irritant
3-Isobutyl-1-methylxanthine
Catalog # SERA26445.02
Supplier:  Serva
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :222.25 g/mol
Melting Pt:200…201 °C
EINECS:249-259-3
 :GHS07
CAS Number:28822-58-4
Formula:C₁₀H₁₄N₄O₂
MDL Number:MFCD00005584