Order Entry
Belgium
Orders LinkContactUsLinkComponent
Hypoxanthine

Hypoxanthine

Marketsource
Supplier:   Serva
Hypoxanthine
Supplier:  Serva
Marketsource
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  1H-Purin-6-one, 1,9-Dihydro-6H-purin-6-one, Purin-6(1H)-one, 6-Purinone
  • CAS Number:68-94-0
  • Formula:C₅H₄N₄O
  • MDL Number:MFCD00005725
  • Molecular Weight:136.11 g/mol
  • Merck Index:13,04895
  • Melting Pt:> 310 °C
  • EINECS:200-697-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g

    Specifications:
    Marketsource
    Marketsource