Order Entry
Belgium
Orders LinkContactUsLinkComponent
COD standard solutions

COD standard solutions

Supplier:   WTW
COD standard solutions
Supplier:  WTW
Certificates
COD standard solutions

2 Options Available Below

Product Details & Documents

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • WTWA252450
  • WTWA252451
  • Physical form:
  • Liquid
  • Liquid

Specifications

Product Family Options

Product Information

  • Concentration
    Pk
    Availability
    Price
  • WTWA252450
    COD standard solution, volume: 100 ml, model: SL COD 100
    COD standard solution, volume: 100 ml, model: SL COD 100
    100 mg/l
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 55,00 €
    55,00 €
    1 * 1 ST
       55,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    WTWA252450
    Physical form
    Liquid
  • WTWA252451
    COD standard solution, volume: 30 ml, model: SL COD 400
    COD standard solution, volume: 30 ml, model: SL COD 400
    400 mg/l
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 55,00 €
    55,00 €
    1 * 1 ST
       55,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    WTWA252451
    Physical form
    Liquid