Order Entry
Belgium
ContactUsLinkComponent
Mouse Recombinant IL-11 (from E. coli)
Mouse Recombinant IL-11 (from E. coli)
Certificates
undefined
Mouse Recombinant IL-11 (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological activity:
    Bioactivity assay: B9 cell proliferation, ED50 ≤250 ng/ml, bioactivity ≥4,0×10³ units/mg
  • Gene ID:
    P47873
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    oprelvekin|AGIF (Adipogenesis Inhibitory Factor)
  • UniProtKB:
    P47873
  • Protein/peptide name:
    IL-11
  • Purity:
    ≥95%
  • Molecular weight:
    19,3 kDa
  • Sequence:
    MPGPPAGSPR VSSDPRADLD SAVLLTRSLL ADTRQLAAQM RDKFPADGDH SLDSLPTLAM SAGTLGSLQL PGVLTRLRVD LMSYLRHVQW LRRAGGPSLK TLEPELGALQ ARLERLLRRL QLLMSRLALP QAAPDQPVIP LGPPASAWGS IRAAHAILGG LHLTLDWAVR GLLLLKTRL
  • Endotoxin level:
    Low
  • Formulation:
    0,1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Frequently Bought Together