Mouse Recombinant MCP-2 (from E. coli)
Supplier: Shenandoah Biotechnology
About this item
Monocyte chemotactic protein 2 (MCP-2), also known as CCL8, is a cytokine that is important during allergic and inflammatory responses. MCP-2 activates mast cells, eosinophils, and basophils through the G protein-coupled chemokine receptors CCR1, CCR2B, and CCR5.
- High quality
- Low endotoxin
- Affordable
: For research use only.
5 Options Available Below
Product Details & Documents
Monocyte chemotactic protein 2 (MCP-2), also known as CCL8, is a cytokine that is important during allergic and inflammatory responses. MCP-2 activates mast cells, eosinophils, and basophils through the G protein-coupled chemokine receptors CCR1, CCR2B, and CCR5.
- High quality
- Low endotoxin
- Affordable
: For research use only.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- SHBT200-63-5UG
- SHBT200-63-20UG
- SHBT200-63-100UG
- SHBT200-63-500UG
- SHBT200-63-1MG
- Protein/peptide name:
- MCP-2
- MCP-2
- MCP-2
- MCP-2
- MCP-2
- Protein synonyms:
- HC14|CCL8
- HC14|CCL8
- HC14|CCL8
- HC14|CCL8
- HC14|CCL8
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- Q9Z121
- Q9Z121
- Q9Z121
- Q9Z121
- Q9Z121
- Sequence:
- GPDKAPVTCC FHVLKLKIPL RVLKSYERIN NIQCPMEAVV FQTKQGMSLC VDPTQKWVSE YMEILDQKSQ ILQP
- GPDKAPVTCC FHVLKLKIPL RVLKSYERIN NIQCPMEAVV FQTKQGMSLC VDPTQKWVSE YMEILDQKSQ ILQP
- GPDKAPVTCC FHVLKLKIPL RVLKSYERIN NIQCPMEAVV FQTKQGMSLC VDPTQKWVSE YMEILDQKSQ ILQP
- GPDKAPVTCC FHVLKLKIPL RVLKSYERIN NIQCPMEAVV FQTKQGMSLC VDPTQKWVSE YMEILDQKSQ ILQP
- GPDKAPVTCC FHVLKLKIPL RVLKSYERIN NIQCPMEAVV FQTKQGMSLC VDPTQKWVSE YMEILDQKSQ ILQP
- Protease-free:
- N
- N
- N
- N
- N
- Animal-free:
- N
- N
- N
- N
- N
- Nuclease-free:
- N
- N
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- N
- Endotoxin level:
- Low
- Low
- Low
- Low
- Low
- Endotoxin content:
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Purity:
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- Grade:
- RUO
- RUO
- RUO
- RUO
- RUO
- Formulation:
- 0,1% Trifluoroacetic acid (TFA)
- 0,1% Trifluoroacetic acid (TFA)
- 0,1% Trifluoroacetic acid (TFA)
- 0,1% Trifluoroacetic acid (TFA)
- 0,1% Trifluoroacetic acid (TFA)
- Molecular weight:
- 8,5 kDa
- 8,5 kDa
- 8,5 kDa
- 8,5 kDa
- 8,5 kDa
- Reconstitution instructions:
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Storage conditions:
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
- RT
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.SHBT200-63-5UGProtein/peptide nameMCP-2Protein synonymsHC14|CCL8Protein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDQ9Z121SequenceGPDKAPVTCC FHVLKLKIPL RVLKSYERIN NIQCPMEAVV FQTKQGMSLC VDPTQKWVSE YMEILDQKSQ ILQPProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic acid (TFA)Molecular weight8,5 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRT - Specifications:
More
Specifications
Cat. No.SHBT200-63-20UGProtein/peptide nameMCP-2Protein synonymsHC14|CCL8Protein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDQ9Z121SequenceGPDKAPVTCC FHVLKLKIPL RVLKSYERIN NIQCPMEAVV FQTKQGMSLC VDPTQKWVSE YMEILDQKSQ ILQPProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic acid (TFA)Molecular weight8,5 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRT - Specifications:
More
Specifications
Cat. No.SHBT200-63-100UGProtein/peptide nameMCP-2Protein synonymsHC14|CCL8Protein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDQ9Z121SequenceGPDKAPVTCC FHVLKLKIPL RVLKSYERIN NIQCPMEAVV FQTKQGMSLC VDPTQKWVSE YMEILDQKSQ ILQPProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic acid (TFA)Molecular weight8,5 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRT - Specifications:
More
Specifications
Cat. No.SHBT200-63-500UGProtein/peptide nameMCP-2Protein synonymsHC14|CCL8Protein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDQ9Z121SequenceGPDKAPVTCC FHVLKLKIPL RVLKSYERIN NIQCPMEAVV FQTKQGMSLC VDPTQKWVSE YMEILDQKSQ ILQPProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic acid (TFA)Molecular weight8,5 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRT - Specifications:
More
Specifications
Cat. No.SHBT200-63-1MGProtein/peptide nameMCP-2Protein synonymsHC14|CCL8Protein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDQ9Z121SequenceGPDKAPVTCC FHVLKLKIPL RVLKSYERIN NIQCPMEAVV FQTKQGMSLC VDPTQKWVSE YMEILDQKSQ ILQPProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic acid (TFA)Molecular weight8,5 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRT



