Human Recombinant MCP-2 (from E. coli)
About this item
Monocyte chemotactic protein 2 (MCP-2), also known as CCL8, is a cytokine that is important during allergic and inflammatory responses. MCP-2 activates mast cells, eosinophils, and basophils through the G protein-coupled chemokine receptors CCR1, CCR2B, and CCR5. MCP-2 signaling through the CCR5 receptor also functions as a natural inhibitor of the human immunodeficiency virus, type 1 (HIV-1).
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
4 Options Available Below
Product Details & Documents
Monocyte chemotactic protein 2 (MCP-2), also known as CCL8, is a cytokine that is important during allergic and inflammatory responses. MCP-2 activates mast cells, eosinophils, and basophils through the G protein-coupled chemokine receptors CCR1, CCR2B, and CCR5. MCP-2 signaling through the CCR5 receptor also functions as a natural inhibitor of the human immunodeficiency virus, type 1 (HIV-1).
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- SHBT100-130-10UG
- SHBT100-130-100UG
- SHBT100-130-500UG
- SHBT100-130-1MG
- Protein/peptide name:
- MCP-2
- MCP-2
- MCP-2
- MCP-2
- Protein synonyms:
- HC14|CCL8
- HC14|CCL8
- HC14|CCL8
- HC14|CCL8
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Human
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- P80075
- P80075
- P80075
- P80075
- Sequence:
- QPDSVSIPIT CCFNVINRKI PIQRLESYTR ITNIQCPKEA VIFKTKRGKE VCADPKERWV RDSMKHLDQI FQNLKP
- QPDSVSIPIT CCFNVINRKI PIQRLESYTR ITNIQCPKEA VIFKTKRGKE VCADPKERWV RDSMKHLDQI FQNLKP
- QPDSVSIPIT CCFNVINRKI PIQRLESYTR ITNIQCPKEA VIFKTKRGKE VCADPKERWV RDSMKHLDQI FQNLKP
- QPDSVSIPIT CCFNVINRKI PIQRLESYTR ITNIQCPKEA VIFKTKRGKE VCADPKERWV RDSMKHLDQI FQNLKP
- Protease-free:
- N
- N
- N
- N
- Animal-free:
- N
- N
- N
- N
- Nuclease-free:
- N
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- Endotoxin level:
- Low
- Low
- Low
- Low
- Endotoxin content:
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Purity:
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- Grade:
- RUO
- RUO
- RUO
- RUO
- Formulation:
- 0,1% Trifluoroacetic acid (TFA)
- 0,1% Trifluoroacetic acid (TFA)
- 0,1% Trifluoroacetic acid (TFA)
- 0,1% Trifluoroacetic acid (TFA)
- Molecular weight:
- 8,9 kDa
- 8,9 kDa
- 8,9 kDa
- 8,9 kDa
- Reconstitution instructions:
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Storage conditions:
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.SHBT100-130-10UGProtein/peptide nameMCP-2Protein synonymsHC14|CCL8Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP80075SequenceQPDSVSIPIT CCFNVINRKI PIQRLESYTR ITNIQCPKEA VIFKTKRGKE VCADPKERWV RDSMKHLDQI FQNLKPProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic acid (TFA)Molecular weight8,9 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRT - Specifications:
More
Specifications
Cat. No.SHBT100-130-100UGProtein/peptide nameMCP-2Protein synonymsHC14|CCL8Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP80075SequenceQPDSVSIPIT CCFNVINRKI PIQRLESYTR ITNIQCPKEA VIFKTKRGKE VCADPKERWV RDSMKHLDQI FQNLKPProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic acid (TFA)Molecular weight8,9 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRT - Specifications:
More
Specifications
Cat. No.SHBT100-130-500UGProtein/peptide nameMCP-2Protein synonymsHC14|CCL8Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP80075SequenceQPDSVSIPIT CCFNVINRKI PIQRLESYTR ITNIQCPKEA VIFKTKRGKE VCADPKERWV RDSMKHLDQI FQNLKPProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic acid (TFA)Molecular weight8,9 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRT - Specifications:
More
Specifications
Cat. No.SHBT100-130-1MGProtein/peptide nameMCP-2Protein synonymsHC14|CCL8Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP80075SequenceQPDSVSIPIT CCFNVINRKI PIQRLESYTR ITNIQCPKEA VIFKTKRGKE VCADPKERWV RDSMKHLDQI FQNLKPProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic acid (TFA)Molecular weight8,9 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRT
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



