Mouse Recombinant IL-17AF (from E. coli)
About this item
Interleukin 11 (IL-11) is a member of the gp130 family of cytokines. IL-11 functions to promote hematopoietic stem cell proliferation and megakaryocyte differentiation. In non-hematopoietic cell populations, IL-11 stimulates acute-phase proteins, modulates the development of immunoglobulin-producing B cells, and regulates bone turnover. IL-11 binds the IL-11Rα receptor to activate JAK downstream signaling.
- High quality
- Low endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Delivery information: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Caution: For research use only.
5 Options Available Below
Product Details & Documents
Interleukin 11 (IL-11) is a member of the gp130 family of cytokines. IL-11 functions to promote hematopoietic stem cell proliferation and megakaryocyte differentiation. In non-hematopoietic cell populations, IL-11 stimulates acute-phase proteins, modulates the development of immunoglobulin-producing B cells, and regulates bone turnover. IL-11 binds the IL-11Rα receptor to activate JAK downstream signaling.
- High quality
- Low endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Delivery information: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Caution: For research use only.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- SHBT200-61-5UG
- SHBT200-61-25UG
- SHBT200-61-100UG
- SHBT200-61-500UG
- SHBT200-61-1MG
- Protein/peptide name:
- IL-17AF
- IL-17AF
- IL-17AF
- IL-17AF
- IL-17AF
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- Q62386/Q7TNI7
- Q62386/Q7TNI7
- Q62386/Q7TNI7
- Q62386/Q7TNI7
- Q62386/Q7TNI7
- Sequence:
- IL-17A: MAAIIPQSSA CPNTEAKDFL QNVKVNLKVF NSLGAKVSSR RPSDYLNRST SPWTLHRNED PDRYPSVIWE AQCRHQRCVN AEGKLDHHMN SVLIQQEILV LKREPESCPF TFRVEKMLVG VGCTCVASIV RQAA IL-17F: MRKNPKAGVP ALQKAGNCPP LEDNTVRVDI RIFNQNQGIS VPREFQNRSS SPWDYNITRD PHRFPSEIAE AQCRHSGCIN AQGQEDSTMN SVAIQQEILV LRREPQGCSN SFRLEKMLLK VGCTCVKPIV HQAA
- IL-17A: MAAIIPQSSA CPNTEAKDFL QNVKVNLKVF NSLGAKVSSR RPSDYLNRST SPWTLHRNED PDRYPSVIWE AQCRHQRCVN AEGKLDHHMN SVLIQQEILV LKREPESCPF TFRVEKMLVG VGCTCVASIV RQAA IL-17F: MRKNPKAGVP ALQKAGNCPP LEDNTVRVDI RIFNQNQGIS VPREFQNRSS SPWDYNITRD PHRFPSEIAE AQCRHSGCIN AQGQEDSTMN SVAIQQEILV LRREPQGCSN SFRLEKMLLK VGCTCVKPIV HQAA
- IL-17A: MAAIIPQSSA CPNTEAKDFL QNVKVNLKVF NSLGAKVSSR RPSDYLNRST SPWTLHRNED PDRYPSVIWE AQCRHQRCVN AEGKLDHHMN SVLIQQEILV LKREPESCPF TFRVEKMLVG VGCTCVASIV RQAA IL-17F: MRKNPKAGVP ALQKAGNCPP LEDNTVRVDI RIFNQNQGIS VPREFQNRSS SPWDYNITRD PHRFPSEIAE AQCRHSGCIN AQGQEDSTMN SVAIQQEILV LRREPQGCSN SFRLEKMLLK VGCTCVKPIV HQAA
- IL-17A: MAAIIPQSSA CPNTEAKDFL QNVKVNLKVF NSLGAKVSSR RPSDYLNRST SPWTLHRNED PDRYPSVIWE AQCRHQRCVN AEGKLDHHMN SVLIQQEILV LKREPESCPF TFRVEKMLVG VGCTCVASIV RQAA IL-17F: MRKNPKAGVP ALQKAGNCPP LEDNTVRVDI RIFNQNQGIS VPREFQNRSS SPWDYNITRD PHRFPSEIAE AQCRHSGCIN AQGQEDSTMN SVAIQQEILV LRREPQGCSN SFRLEKMLLK VGCTCVKPIV HQAA
- IL-17A: MAAIIPQSSA CPNTEAKDFL QNVKVNLKVF NSLGAKVSSR RPSDYLNRST SPWTLHRNED PDRYPSVIWE AQCRHQRCVN AEGKLDHHMN SVLIQQEILV LKREPESCPF TFRVEKMLVG VGCTCVASIV RQAA IL-17F: MRKNPKAGVP ALQKAGNCPP LEDNTVRVDI RIFNQNQGIS VPREFQNRSS SPWDYNITRD PHRFPSEIAE AQCRHSGCIN AQGQEDSTMN SVAIQQEILV LRREPQGCSN SFRLEKMLLK VGCTCVKPIV HQAA
- Protease-free:
- N
- N
- N
- N
- N
- Animal-free:
- N
- N
- N
- N
- N
- Nuclease-free:
- N
- N
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- N
- Endotoxin level:
- Low
- Low
- Low
- Low
- Low
- Endotoxin content:
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- UniProtKB:
- Q62386/Q7TNI7
- Q62386/Q7TNI7
- Q62386/Q7TNI7
- Q62386/Q7TNI7
- Q62386/Q7TNI7
- Biological activity:
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Purity:
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- Grade:
- RUO
- RUO
- RUO
- RUO
- RUO
- Formulation:
- 0,1% Trifluoroacetic Acid (TFA)
- 0,1% Trifluoroacetic Acid (TFA)
- 0,1% Trifluoroacetic Acid (TFA)
- 0,1% Trifluoroacetic Acid (TFA)
- 0,1% Trifluoroacetic Acid (TFA)
- Molecular weight:
- 30,1 kDa
- 30,1 kDa
- 30,1 kDa
- 30,1 kDa
- 30,1 kDa
- Reconstitution instructions:
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Storage conditions:
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
- RT
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.SHBT200-61-5UGProtein/peptide nameIL-17AFProtein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDQ62386/Q7TNI7SequenceIL-17A: MAAIIPQSSA CPNTEAKDFL QNVKVNLKVF NSLGAKVSSR RPSDYLNRST SPWTLHRNED PDRYPSVIWE AQCRHQRCVN AEGKLDHHMN SVLIQQEILV LKREPESCPF TFRVEKMLVG VGCTCVASIV RQAA IL-17F: MRKNPKAGVP ALQKAGNCPP LEDNTVRVDI RIFNQNQGIS VPREFQNRSS SPWDYNITRD PHRFPSEIAE AQCRHSGCIN AQGQEDSTMN SVAIQQEILV LRREPQGCSN SFRLEKMLLK VGCTCVKPIV HQAAProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1UniProtKBQ62386/Q7TNI7Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight30,1 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRT - Specifications:
More
Specifications
Cat. No.SHBT200-61-25UGProtein/peptide nameIL-17AFProtein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDQ62386/Q7TNI7SequenceIL-17A: MAAIIPQSSA CPNTEAKDFL QNVKVNLKVF NSLGAKVSSR RPSDYLNRST SPWTLHRNED PDRYPSVIWE AQCRHQRCVN AEGKLDHHMN SVLIQQEILV LKREPESCPF TFRVEKMLVG VGCTCVASIV RQAA IL-17F: MRKNPKAGVP ALQKAGNCPP LEDNTVRVDI RIFNQNQGIS VPREFQNRSS SPWDYNITRD PHRFPSEIAE AQCRHSGCIN AQGQEDSTMN SVAIQQEILV LRREPQGCSN SFRLEKMLLK VGCTCVKPIV HQAAProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1UniProtKBQ62386/Q7TNI7Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight30,1 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRT - Specifications:
More
Specifications
Cat. No.SHBT200-61-100UGProtein/peptide nameIL-17AFProtein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDQ62386/Q7TNI7SequenceIL-17A: MAAIIPQSSA CPNTEAKDFL QNVKVNLKVF NSLGAKVSSR RPSDYLNRST SPWTLHRNED PDRYPSVIWE AQCRHQRCVN AEGKLDHHMN SVLIQQEILV LKREPESCPF TFRVEKMLVG VGCTCVASIV RQAA IL-17F: MRKNPKAGVP ALQKAGNCPP LEDNTVRVDI RIFNQNQGIS VPREFQNRSS SPWDYNITRD PHRFPSEIAE AQCRHSGCIN AQGQEDSTMN SVAIQQEILV LRREPQGCSN SFRLEKMLLK VGCTCVKPIV HQAAProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1UniProtKBQ62386/Q7TNI7Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight30,1 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRT - Specifications:
More
Specifications
Cat. No.SHBT200-61-500UGProtein/peptide nameIL-17AFProtein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDQ62386/Q7TNI7SequenceIL-17A: MAAIIPQSSA CPNTEAKDFL QNVKVNLKVF NSLGAKVSSR RPSDYLNRST SPWTLHRNED PDRYPSVIWE AQCRHQRCVN AEGKLDHHMN SVLIQQEILV LKREPESCPF TFRVEKMLVG VGCTCVASIV RQAA IL-17F: MRKNPKAGVP ALQKAGNCPP LEDNTVRVDI RIFNQNQGIS VPREFQNRSS SPWDYNITRD PHRFPSEIAE AQCRHSGCIN AQGQEDSTMN SVAIQQEILV LRREPQGCSN SFRLEKMLLK VGCTCVKPIV HQAAProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1UniProtKBQ62386/Q7TNI7Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight30,1 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRT - Specifications:
More
Specifications
Cat. No.SHBT200-61-1MGProtein/peptide nameIL-17AFProtein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDQ62386/Q7TNI7SequenceIL-17A: MAAIIPQSSA CPNTEAKDFL QNVKVNLKVF NSLGAKVSSR RPSDYLNRST SPWTLHRNED PDRYPSVIWE AQCRHQRCVN AEGKLDHHMN SVLIQQEILV LKREPESCPF TFRVEKMLVG VGCTCVASIV RQAA IL-17F: MRKNPKAGVP ALQKAGNCPP LEDNTVRVDI RIFNQNQGIS VPREFQNRSS SPWDYNITRD PHRFPSEIAE AQCRHSGCIN AQGQEDSTMN SVAIQQEILV LRREPQGCSN SFRLEKMLLK VGCTCVKPIV HQAAProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1UniProtKBQ62386/Q7TNI7Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight30,1 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRT



