Order Entry
Belgium
Orders LinkContactUsLinkComponent
Human recombinant Eotaxin (from <i>E. coli</i>)

Human recombinant Eotaxin (from E. coli)

Human recombinant Eotaxin (from <i>E. coli</i>)
Certificates
Human recombinant Eotaxin (from <i>E. coli</i>)

5 Options Available Below

Product Details & Documents

Eotaxin, also known as Eotaxin-1 and CCL11, is a chemokine that recruits eosinophils to sites of allergic inflammation. Eotaxin signals through the G protein-coupled chemokine receptors CCR2, CCR3, and CCR5.

  • Produced from genetically modified E. coli
  • Monomeric protein, containing 74 amino acids

Eotaxin also negatively regulates adult neurogenesis, and high Eotaxin levels are associated with impaired learning and memory.

Delivery information: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.

Caution: This product is for research use only and is not for use in humans.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SHBT100-178-5UG
  • SHBT100-178-20UG
  • SHBT100-178-100UG
  • SHBT100-178-500UG
  • SHBT100-178-1MG
  • Protein/peptide name:
  • Eotaxin
  • Eotaxin
  • Eotaxin
  • Eotaxin
  • Eotaxin
  • Protein synonyms:
  • Eosinophil chemotactic protein|CCL11
  • Eosinophil chemotactic protein|CCL11
  • Eosinophil chemotactic protein|CCL11
  • Eosinophil chemotactic protein|CCL11
  • Eosinophil chemotactic protein|CCL11
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Human
  • Human
  • Human
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • P51671
  • P51671
  • P51671
  • P51671
  • P51671
  • Sequence:
  • GPASVPTTCC FNLANRKIPL QRLESYRRIT SGKCPQKAVI FKTKLAKDIC ADPKKKWVQD SMKYLDQKSP TPKP
  • GPASVPTTCC FNLANRKIPL QRLESYRRIT SGKCPQKAVI FKTKLAKDIC ADPKKKWVQD SMKYLDQKSP TPKP
  • GPASVPTTCC FNLANRKIPL QRLESYRRIT SGKCPQKAVI FKTKLAKDIC ADPKKKWVQD SMKYLDQKSP TPKP
  • GPASVPTTCC FNLANRKIPL QRLESYRRIT SGKCPQKAVI FKTKLAKDIC ADPKKKWVQD SMKYLDQKSP TPKP
  • GPASVPTTCC FNLANRKIPL QRLESYRRIT SGKCPQKAVI FKTKLAKDIC ADPKKKWVQD SMKYLDQKSP TPKP
  • Protease-free:
  • N
  • N
  • N
  • N
  • N
  • Animal-free:
  • N
  • N
  • N
  • N
  • N
  • Nuclease-free:
  • N
  • N
  • N
  • N
  • N
  • Carrier-free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • N
  • N
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • Grade:
  • RUO
  • RUO
  • RUO
  • RUO
  • RUO
  • Formulation:
  • 0,1% Trifluoroacetic Acid (TFA)
  • 0,1% Trifluoroacetic Acid (TFA)
  • 0,1% Trifluoroacetic Acid (TFA)
  • 0,1% Trifluoroacetic Acid (TFA)
  • 0,1% Trifluoroacetic Acid (TFA)
  • Molecular weight:
  • 8,4 kDa
  • 8,4 kDa
  • 8,4 kDa
  • 8,4 kDa
  • 8,4 kDa
  • Reconstitution instructions:
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Storage conditions:
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • Shipping temperature:
  • RT
  • RT
  • RT
  • RT
  • RT

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Vial
    5 µG
    Log in for availability
    Log in for availability
    1 * 5 µG
    1 * 5 µG - 76,50 €
    76,50 €
    1 * 5 µG
       76,50 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-178-5UG
    Protein/peptide name
    Eotaxin
    Protein synonyms
    Eosinophil chemotactic protein|CCL11
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P51671
    Sequence
    GPASVPTTCC FNLANRKIPL QRLESYRRIT SGKCPQKAVI FKTKLAKDIC ADPKKKWVQD SMKYLDQKSP TPKP
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA)
    Molecular weight
    8,4 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
  • Vial
    20 µG
    Log in for availability
    Log in for availability
    1 * 20 µG
    1 * 20 µG - 190,00 €
    190,00 €
    1 * 20 µG
       190,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-178-20UG
    Protein/peptide name
    Eotaxin
    Protein synonyms
    Eosinophil chemotactic protein|CCL11
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P51671
    Sequence
    GPASVPTTCC FNLANRKIPL QRLESYRRIT SGKCPQKAVI FKTKLAKDIC ADPKKKWVQD SMKYLDQKSP TPKP
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA)
    Molecular weight
    8,4 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
  • Vial
    100 µG
    Log in for availability
    Log in for availability
    1 * 1 EA
    1 * 1 EA - 476,00 €
    476,00 €
    1 * 1 EA
       476,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-178-100UG
    Protein/peptide name
    Eotaxin
    Protein synonyms
    Eosinophil chemotactic protein|CCL11
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P51671
    Sequence
    GPASVPTTCC FNLANRKIPL QRLESYRRIT SGKCPQKAVI FKTKLAKDIC ADPKKKWVQD SMKYLDQKSP TPKP
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA)
    Molecular weight
    8,4 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
  • Vial
    500 µG
    Log in for availability
    Log in for availability
    1 * 500 µG
    1 * 500 µG - 2 060,00 €
    2 060,00 €
    1 * 500 µG
       2 060,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-178-500UG
    Protein/peptide name
    Eotaxin
    Protein synonyms
    Eosinophil chemotactic protein|CCL11
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P51671
    Sequence
    GPASVPTTCC FNLANRKIPL QRLESYRRIT SGKCPQKAVI FKTKLAKDIC ADPKKKWVQD SMKYLDQKSP TPKP
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA)
    Molecular weight
    8,4 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
  • Vial
    1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 3 420,00 €
    3 420,00 €
    1 * 1 mg
       3 420,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-178-1MG
    Protein/peptide name
    Eotaxin
    Protein synonyms
    Eosinophil chemotactic protein|CCL11
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P51671
    Sequence
    GPASVPTTCC FNLANRKIPL QRLESYRRIT SGKCPQKAVI FKTKLAKDIC ADPKKKWVQD SMKYLDQKSP TPKP
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA)
    Molecular weight
    8,4 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.