Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ethidium bromide

Ethidium bromide

Glass bottle for solids, 25 g
Catalog # MOLE34277542-25G
Toxic
Health
Marketsource
Supplier:   Molekula
Ethidium bromide
Catalog # MOLE34277542-25G
Supplier:  Molekula
Marketsource
CAS Number:  1239-45-8
Ethidium bromide

Glass bottle for solids, 25 g

CAS Number:  1239-45-8
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
  • CAS Number:1239-45-8
  • Formula:C₂₁H₂₀BrN₃
  • MDL Number:MFCD00011724
  • Molecular Weight:394.31 g/mol
  • Merck Index:13,04751
  • Boiling Pt:72 °C (0.1 mmHg)
  • Melting Pt:261…264 °C
  • Density:1.01 g/cm³ (20 °C)
  • Flash Pt:<100 °C
  • Storage Temperature:Ambient
  • EINECS:214-984-6
  • UN:2811
  • ADR:6.1,I
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.