Ethidium bromide
Glass bottle for solids, 5 g
Catalog # MOLEM16559763
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
- CAS Number:1239-45-8
- Formula:C₂₁H₂₀BrN₃
- MDL Number:MFCD00011724
- Molecular Weight:394.31 g/mol
- Merck Index:13,04751
- Boiling Pt:72 °C (0.1 mmHg)
- Melting Pt:261…264 °C
- Density:1.01 g/cm³ (20 °C)
- Flash Pt:<100 °C
- Storage Temperature:Ambient
- EINECS:214-984-6
- UN:2811
- ADR:6.1,I
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



