Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ethidium bromide 95%, pure

Ethidium bromide 95%, pure

Toxic
Health
Ethidium bromide 95%, pure
Certificates
Ethidium bromide 95%, pure

3 Options Available Below

Product Details & Documents
Synonyms:  3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
  • CAS Number:1239-45-8
  • Formula:C₂₁H₂₀BrN₃
  • MDL Number:MFCD00011724
  • Molecular Weight:394.31 g/mol
  • Merck Index:13,04751
  • Boiling Pt:72 °C (0,1 -rr)
  • Melting Pt:260…262 °C (dec.)
  • Density:1,01 g/cm³ (20 °C)
  • Flash Pt:>100 °C
  • Storage Temperature:Ambient
  • EINECS:214-984-6
  • UN:2811
  • ADR:6.1,I

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Red to dark purple Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    ≥94 %
  • Loss on drying:
    ≤5.0 %
  • Solubility:
    (1 % in water) Clear red
  • pH:
    (1 % in water at 20°C) Approx. 4.5
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 56,80 €
    56,80 €
    1 * 1 g
       56,80 €

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 186,00 €
    186,00 €
    1 * 5 g
       186,00 €

  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 714,00 €
    714,00 €
    1 * 25 g
       714,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.