Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sodium L(+)-ascorbate

Sodium L(+)-ascorbate

Glass bottle for solids, 50 mg
Catalog # EDQMY0000039
Supplier:   EDQM
Sodium L(+)-ascorbate
Catalog # EDQMY0000039
Supplier:  EDQM
CAS Number:  134-03-2
Sodium L(+)-ascorbate
Sodium L(+)-ascorbate

Glass bottle for solids, 50 mg

CAS Number:  134-03-2
Price
270,00 €
Log in for availability
1 * 50 mg
Product Details & Documents
Synonyms:  Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt
  • CAS Number:134-03-2
  • Formula:C₆H₇NaO₆
  • MDL Number:MFCD00082340
  • Molecular Weight:198.11 g/mol
  • Melting Pt:219 °C
  • Density:1.799 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:205-126-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.