Acide L(+)-ascorbique 99%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- CAS Number:50-81-7
- Formula:C₆H₈O₆
- MDL Number:MFCD00064328
- Molecular Weight:176.13 g/mol
- Merck Index:13,00837
- Boiling Pt:553 °C (1013 hPa)
- Melting Pt:190…192 °C (déc.)
- Density:1,694 g/cm³ (20 °C)
- Storage Temperature:Température ambiante
- EINECS:200-066-2
- REACH:exempt
3 Options Available Below
Product Details & Documents
Synonyms: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- CAS Number:50-81-7
- Formula:C₆H₈O₆
- MDL Number:MFCD00064328
- Molecular Weight:176.13 g/mol
- Merck Index:13,00837
- Boiling Pt:553 °C (1013 hPa)
- Melting Pt:190…192 °C (déc.)
- Density:1,694 g/cm³ (20 °C)
- Storage Temperature:Température ambiante
- EINECS:200-066-2
- REACH:exempt
Specifications for Product Family
Specification Test Results
- Appearance:White to off-white Crystalline powder or crystals
- Infrared spectrum:Conforms
- Titration Iodimetric:≥98.5 %
- Sulfated ash:≤0.1 %
- Solubility:(5 % in water) Passes test
- pH:2.1 to 2.6 (5 % in water)
- Specific optical rotation:+20.5° to +21.5° (20°C, 589 nm) (c=10, H2O)
- Copper (Cu):≤5 ppm
- Iron (Fe):≤2 ppm
- Heavy metals (as Pb):≤10 ppm
- Impurity:≤0.2 % Oxalic acid
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



