Spironolactone 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:52-01-7
:
Formula:C₂₄H₃₂O₄S
MDL Number:MFCD00082250
Merck Index:12,08917
Boiling Pt:597 °C (1013 hPa)
Melting Pt:198…207 °C
:416.58 g/mol
Storage Temperature:Ambient
EINECS:200-133-6
:GHS08
Specification Test Results
- Appearance:White to yellowish-white Powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (105°C, 2 h)
- HPLC:≥98.5 %
- Specific optical rotation:-41° to -45° (20°C, 589 nm) (c=1, EtOH)
Specification Test Results
Frequently Bought Together