


Palladium(II) chloride, anhydrous for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7647-10-1
:
Formula:PdCl₂
MDL Number:MFCD00003558
Merck Index:13,07058
Melting Pt:678 °C
Density:4 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:8,II
:M-Clarity™ quality level = MQ200
:177.33 g/mol
EINECS:231-596-2
UN:3260
:GHS05
:GHS07
:GHS09
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:680 °C
- Molar Mass:177.33 g/mol
- Density:4.0 g/cm3 (20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
Related Information
Frequently Bought Together